Avantor® Hichrom HI-WAX MS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI25-20-100-60 HI25-32-300-10 HI25-10-010-15 HI25-10-010-10 HI25-10-040-10 HI25-10-020-5 HI25-53-010-10 HI25-20-040-50 HI25-25-010-30 HI252502515G5 HI25-20-100-50 HI25-32-010-10 HI25-32-025-30 HI25-20-100-10 HI25-53-100-60 HI25-18-018-60 HI25-53-025-60 HI25-53-050-30 HI25-25-025-10 HI25-32-050-30 HI25050052.5 HI25-18-018-50 HI25-25-100-30 HI25-53-250-15 HI25-32-100-15 HI25-53-250-10 HI25-15-015-10 HI25-05-005-5 HI25-32-025-15 HI25-53-200-10 HI25-53-150-60 HI25-53-100-15 HI25-25-100-60 HI25-32-050-50 HI25-32-025-10 HI25-10-020-15 HI2532100105 HI25-53-100-50 HI25-32-050-10 HI25-53-200-60 HI25-25-010-15 HI25-18-020-50 HI25-53-010-30 HI25-05-005-2 HI25-32-100-60 HI25-20-010-20 HI25-10-020-10 HI25-53-100-75 HI25-32-010-50 HI25-32-150-15 HI25-25-010-50 HI2532150105 HI252502530G5 HI25-20-010-10 HI25-32-300-50 HI25-18-020-10 HI25-18-020-60 HI25-18-018-10 HI2532300105 HI25-53-050-10 HI25-20-040-60 HI25-53-025-15 HI25-25-050-50 HI25-53-025-50 HI25-32-010-30 HI2532010105 HI25-15-015-20 HI25-25-025-30 HI25-18-040-10 HI25-32-025-60 HI25-53-150-50 HI25-53-050-15 HI25-15-015-15 HI25-32-010-60 HI25-32-150-10 HI25-18-040-60 HI25-32-150-30 HI25-25-025-60 HI25-10-040-15 HI25-53-050-50 HI25-53-100-30 HI25-53-250-60 HI25-15-015-5 HI25-53-050-60 HI25-25-050-10 HI25-20-010-50 HI25-53-010-15 HI25-32-050-15 HI25-20-040-10 HI25-53-200-50 HI25-32-300-30 HI25-20-020-40 HI25-32-010-15 HI2525025105 HI25-10-010-5 HI25-25-100-15 HI25-53-250-75 HI25-32-300-15 HI25-20-010-40 HI25-32-150-60 HI25-20-040-40 HI25-53-250-50 HI25-20-010-60 HI25-53-200-15 HI25-53-150-30 HI25-18-018-40 HI25-25-050-60 HI25-25-025-15 HI25-20-040-20 HI25-32-050-60 HI25-18-020-20 HI25-53-200-30 HI25-20-020-50 HI25-53-150-15 HI25-25-025-50 HI2532025105 HI25-25-010-60 HI25-53-100-10 HI25-53-150-75 HI2525050105 HI25-20-020-10 HI25-25-050-15 HI25-53-250-30 HI25-32-100-30 HI25-10-040-5 HI255310030G5 HI25-32-100-50 HI25-32-100-10 HI25-18-018-20 HI25-53-150-10 HI25-25-050-30 HI25-15-015-30 HI25-18-040-50 HI25-25-100-10 HI2525100105 HI25-53-200-75 HI2532050105 HI25-20-020-20 HI25-32-300-60 HI25-53-025-30 HI252502560G5 HI25-18-040-20 HI25-20-100-20 HI25-20-100-40 HI25-18-040-40 HI25-32-150-50 HI25-18-020-40 HI25-25-100-50 HI25-20-020-60 HI25-32-025-50
76433-814EA 1894.4 USD
76433-814 76433-778 76433-812 76433-658 76433-818 76433-816 76433-780 76433-662 76433-784 76433-660 76433-782 76433-666 76433-820 76433-700 76433-788 76433-664 76433-786 76433-802 76433-648 76433-800 76433-646 76433-768 76433-806 76433-804 76433-808 76433-772 76433-652 76433-770 76433-650 76433-776 76433-810 76433-656 76433-774 76433-654 76433-758 76433-638 76433-756 76433-636 76433-640 76433-762 76433-760 76433-644 76433-766 76433-642 76433-764 76433-626 76433-748 76433-624 76433-746 76434-150 76433-628 76434-152 76433-750 76433-596 76433-630 76433-594 76433-754 76433-634 76433-598 76433-752 76433-632 76433-736 76433-616 76433-734 76433-614 76433-738 76434-140 76433-618 76434-142 76434-144 76434-146 76434-148 76433-740 76433-622 76433-744 76433-620 76433-742 76433-604 76433-846 76433-726 76433-602 76433-844 76433-724 76433-608 76433-848 76433-606 76434-130 76433-728 76434-132 76434-134 76434-136 76434-138 76434-128 76433-692 76433-690 76433-850 76433-696 76433-694 76433-732 76433-612 76433-730 76433-610 76433-698 76433-714 76433-836 76433-712 76433-834 76433-718 76433-716 76433-838 76434-120 76434-122 76434-124 76434-126 76433-680 76433-684 76433-682 76433-600 76433-842 76433-688 76433-722 76433-840 76433-686 76433-720 76433-824 76433-704 76433-668 76433-822 76433-702 76433-828 76433-708 76433-826 76433-706 76431-440 76433-790 76433-670 76433-794 76433-674 76433-792 76431-446 76433-672 76433-798 76433-710 76431-444 76433-678 76433-832 76431-442 76433-796 76433-830 76433-676
Avantor® Hichrom HI-WAX MS GC Columns
Chromatography Columns
Very popular PEG high polarity MS phase for a wide range of applications, including (for example): essential oils, flavors and fragrances, alkenols. HI-WAX MS is an MS grade low-bleeding crossbonded column.
  • Phase composition: Crossbonded Polyethylene Glycol (PEG) - MS grade
  • Long lifetime
Maximum temperature limits may change depending on the film thickness.