Avantor® Hichrom HI-624 MS, GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI24-18-100-40 HI245330030G5 HI24-25-100-30 HI24-32-010-30 HI24-18-018-50 HI2432150105 HI242514030G5 HI24-53-300-50 HI24-32-050-60 HI24-32-300-10 HI24-25-050-10 HI2432100105 HI243218030G5 HI24-25-300-50 HI24-15-015-5 HI24-15-015-20 HI24-32-010-15 HI24050052.5 HI24-53-025-15 HI24-32-300-60 HI24-18-040-50 HI24-25-010-60 HI24-18-018-60 HI24-53-100-15 HI24-18-020-60 HI24-18-100-60 HI24-53-010-10 HI24-53-100-75 HI24-05-005-2 HI24-53-150-10 HI24-25-025-15 HI242514060G5 HI24-25-025-60 HI24-18-020-10 HI24-32-180-75 HI24-25-140-30 HI24-10-020-15 HI24-20-112-60 HI24-32-150-50 HI24-18-018-40 HI2432050105 HI24-10-010-10 HI24-53-150-50 HI24-53-050-30 HI24-53-050-50 HI24-32-025-30 HI24-32-025-15 HI24-18-018-20 HI24-25-050-15 HI24-53-150-15 HI24-10-010-15 HI24-53-010-30 HI24-53-300-10 HI24-53-150-30 HI2425140105 HI24-32-100-60 HI24-25-100-50 HI24-53-100-10 HI24-32-180-30 HI24-32-025-10 HI24-32-050-50 HI24-53-300-75 HI24-15-084-15 HI24-25-010-50 HI24-53-050-15 HI24-32-100-15 HI2425050105 HI24-25-010-30 HI24-18-020-20 HI24-18-040-40 HI24-32-300-15 HI24-25-140-60 HI24-25-050-60 HI24-32-010-50 HI242502515G5 HI24-10-040-10 HI24-10-020-10 HI24-53-300-60 HI24-53-050-60 HI24-53-100-50 HI24-32-100-50 HI24-10-040-5 HI24-53-025-50 HI24-15-084-20 HI24-25-100-15 HI24-32-010-10 HI24-25-025-10 HI24-18-100-50 HI24-18-040-10 HI24-20-112-20 HI24-32-300-50 HI2432180105 HI24-53-050-10 HI24-53-150-60 HI242502560G5 HI24-32-180-50 HI24-20-112-50 HI24-32-150-10 HI2432300105 HI24-25-100-10 HI24-05-005-5 HI24-53-100-60 HI24-18-018-10 HI24-15-015-15 HI24-53-150-75 HI24-25-100-60 HI24-53-100-30 HI2425025105 HI245310030G5 HI24-25-140-50 HI24-10-010-5 HI24-32-150-15 HI24-53-300-30 HI2432025105 HI24-10-020-5 HI24-18-100-20 HI24-10-040-15 HI24-25-025-50 HI24-25-300-60 HI24-32-180-60 HI24-32-050-15 HI24-25-300-30 HI24-15-084-40 HI24-32-300-30 HI24-53-025-60 HI24-25-050-30 HI24-32-025-60 HI24-32-100-10 HI2432010105 HI24-32-010-60 HI24-25-010-15 HI24-15-084-30 HI242502530G5 HI24-53-300-15 HI24-32-050-30 HI24-18-040-20 HI24-32-025-50 HI24-15-015-30 HI24-32-150-60 HI24-53-010-15 HI24-18-020-40 HI24-32-100-30 HI24-53-025-30 HI24-32-050-10 HI24-15-015-10 HI24-25-025-30 HI24-20-112-40 HI24-32-150-30 HI24-18-020-50 HI24-25-050-50
76433-382EA 1418.54 USD
76433-382 76431-290 76433-432 76433-458 76433-358 76433-502 76431-282 76433-588 76433-486 76433-522 76433-416 76433-490 76431-286 76433-448 76434-110 76434-106 76433-456 76434-080 76433-540 76433-532 76433-378 76433-402 76433-360 76433-560 76433-370 76433-386 76433-534 76433-568 76434-078 76433-570 76433-408 76431-284 76433-414 76433-362 76433-520 76433-440 76434-092 76433-394 76433-508 76433-356 76433-478 76434-084 76433-576 76433-552 76433-554 76433-470 76433-468 76433-354 76433-420 76433-572 76434-086 76433-538 76433-582 76433-574 76433-438 76433-498 76433-434 76433-558 76433-514 76433-464 76433-484 76433-592 76434-112 76433-400 76433-550 76433-492 76433-418 76433-398 76433-364 76433-376 76433-526 76433-444 76433-426 76433-460 76431-276 76434-096 76434-090 76433-590 76433-556 76433-564 76433-496 76434-100 76433-544 76434-114 76433-430 76433-452 76433-404 76433-384 76433-372 76433-388 76433-530 76433-512 76433-548 76433-578 76431-280 76433-516 76433-392 76433-500 76433-524 76433-428 76434-082 76433-566 76433-352 76434-104 76433-580 76433-436 76433-562 76433-406 76431-288 76433-442 76434-088 76433-504 76433-586 76433-466 76434-094 76433-380 76434-098 76433-412 76433-450 76433-518 76433-480 76433-446 76434-118 76433-528 76433-546 76433-422 76433-474 76433-488 76433-454 76433-462 76433-396 76434-116 76431-278 76433-584 76433-482 76433-374 76433-472 76434-108 76433-510 76433-536 76433-366 76433-494 76433-542 76433-476 76434-102 76433-410 76433-390 76433-506 76433-368 76433-424
Avantor® Hichrom HI-624 MS, GC Columns
Chromatography Columns
General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants. HI-624 MS is an MS grade crossbonded column.
  • Phase composition: Crossbonded 6% cyanopropylphenyl, 94% methyl polysiloxane - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.