Avantor® Hichrom HI-5 MS Plus GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI18-20-020-50 HI18-20-010-60 HI18-10-040-15 HI18-32-050-15 HI18-53-100-75 HI18-18-020-40 HI18-32-025-50 HI18-53-100-50 HI18-53-010-30 HI18-05-005-2 HI18-20-010-20 HI18-32-150-60 HI18-25-050-50 HI18-18-036-60 HI18-20-020-60 HI18-05-005-5 HI18-25-100-10 HI18-25-025-50 HI18-20-033-12 HI18-20-040-10 HI183202530G5 HI182502530G10 HI18-25-100-50 HI18-20-010-40 HI18-20-020-40 HI18-53-100-15 HI18-32-010-10 HI18-18-018-10 HI18-10-010-15 HI18-53-150-60 HI18-20-040-60 HI18-25-050-15 HI18-32-025-10 HI18-25-050-60 HI18-20-100-40 HI185310030G5 HI1825025105 HI18-18-040-60 HI18-53-025-50 HI18-25-100-15 HI18-25-010-30 HI18-53-150-10 HI18-15-015-30 HI18-32-050-50 HI18-15-015-15 HI18-53-150-15 HI18-15-015-5 HI18-10-020-10 HI18-18-020-50 HI18-20-033-25 HI18-32-150-50 HI18-18-040-50 HI18-10-020-15 HI18-53-010-10 HI18-53-025-30 HI18-20-040-50 HI18-20-020-10 HI18-32-100-10 HI1832100105 HI18-15-015-10 HI182502560G10 HI18-32-150-10 HI18-53-050-30 HI18-25-100-30 HI18-18-018-50 HI182505015G10 HI18-20-010-50 HI18-18-040-40 HI18-32-100-60 HI18-53-050-50 HI18-32-100-50 HI18-32-025-30 HI18-25-050-10 HI18-25-010-60 HI182501030G5 HI18-18-040-20 HI1832050105 HI18-18-020-10 HI18-20-033-60 HI18-20-100-60 HI1825100105 HI18-53-100-60 HI18-32-010-30 HI18-25-050-30 HI18-53-050-10 HI18-20-033-50 HI18-20-020-20 HI18-32-050-60 HI18-25-010-15 HI18-25-025-30 HI18-10-020-5 HI18-53-010-15 HI183210030G5 HI18-25-100-60 HI18-32-025-15 HI18050052.5 HI18-20-100-20 HI18-53-050-60 HI18-25-025-10 HI18-53-150-50 HI1832025105 HI18-32-150-15 HI18-32-100-15 HI182505030G5 HI18-53-100-30 HI18-10-040-5 HI18-32-010-50 HI18-10-040-10 HI18-15-015-20 HI18-53-150-75 HI18-20-033-30 HI18-20-010-10 HI18-18-020-60 HI18-20-040-40 HI18-32-010-15 HI18-18-018-40 HI18-53-025-60 HI18-32-150-30 HI18-32-050-30 HI182505030G10 HI18-32-010-60 HI18-32-025-60 HI18-20-100-10 HI18-53-050-15 HI1832010105 HI18-53-150-30 HI18-25-025-60 HI18-20-100-50 HI18-18-036-40 HI18-18-018-60 HI18-18-040-10 HI18-32-050-10 HI18-53-100-10 HI1825050105 HI18-20-040-20 HI18-53-025-15 HI182502560G5 HI18-10-010-10 HI18-25-010-50 HI18-10-010-5 HI18-18-036-20 HI1832150105 HI18-25-025-15 HI18-18-018-20 HI18-32-100-30 HI182502515G5 HI18-18-020-20 HI182502530G5
76432-974EA 734.23 USD
76432-974 76432-976 76432-970 76432-972 76433-020 76432-966 76432-968 76433-024 76433-022 76433-028 76433-026 76433-008 76432-984 76432-986 76432-980 76433-090 76432-982 76433-094 76433-092 76432-978 76433-098 76433-010 76433-096 76433-014 76433-012 76433-018 76433-016 76434-040 76434-042 76432-952 76432-954 76432-950 76432-948 76433-082 76431-232 76433-080 76431-230 76433-086 76432-944 76433-084 76432-946 76433-002 76433-000 76433-088 76433-006 76433-004 76431-234 76433-108 76434-030 76432-962 76434-032 76432-964 76434-034 76434-036 76432-960 76434-038 76433-072 76433-070 76432-956 76433-076 76433-074 76432-958 76433-112 76433-110 76433-078 76432-890 76434-020 76432-896 76432-930 76434-022 76432-898 76432-932 76434-024 76432-892 76434-026 76432-894 76434-028 76432-926 76434-018 76433-060 76432-928 76433-064 76432-922 76433-062 76432-924 76433-068 76433-102 76431-218 76433-066 76433-100 76431-216 76433-106 76431-214 76433-104 76431-212 76434-010 76432-940 76432-942 76434-012 76434-014 76434-016 76431-220 76433-050 76432-938 76432-934 76433-054 76433-052 76432-936 76431-228 76433-058 76431-226 76433-056 76431-224 76431-222 76432-990 76432-996 76432-876 76432-910 76432-998 76432-992 76432-994 76432-904 76432-906 76432-900 76433-042 76432-988 76433-040 76432-902 76433-046 76433-044 76432-908 76433-048 76432-880 76432-886 76432-920 76432-888 76432-882 76432-884 76432-916 76432-918 76432-878 76433-032 76432-912 76432-914 76433-030 76433-036 76433-034 76433-038
Avantor® Hichrom HI-5 MS Plus GC Columns
Chromatography Columns
General purpose GC-MS column with extra stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols, etc.
Maximum temperature limits may change depending on the film thickness.