Avantor® Hichrom HI-5 MS, GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI17-10-040-10 HI17-32-010-30 HI17-18-018-50 HI17-25-100-15 HI1725025105 HI17-18-036-60 HI17-15-015-30 HI17-18-036-40 HI17-25-010-50 HI17-18-018-20 HI175310030G5 HI17-05-005-5 HI1732010105 HI17-32-150-10 HI1732050105 HI17-53-050-10 HI17-20-040-60 HI17-18-018-10 HI17-25-050-15 HI17-32-025-10 HI172505030G5 HI173202530G5 HI17-25-010-30 HI17-32-150-60 HI17-25-050-60 HI17-20-033-12 HI1732025105 HI17-32-010-10 HI17-20-040-50 HI17-32-050-50 HI17-20-033-60 HI172502560G10 HI17-10-020-5 HI17-10-010-15 HI17-32-050-10 HI17-32-150-15 HI173210030G5 HI17-20-010-60 HI17-53-050-50 HI17-25-010-15 HI17-32-100-10 HI17-53-150-15 HI17-18-040-50 HI17-32-050-15 HI17-20-020-20 HI17-25-100-10 HI17-53-010-30 HI17-20-100-40 HI17-53-100-30 HI17-32-025-50 HI17-18-018-40 HI1732150105 HI17-20-020-50 HI17-32-050-60 HI17-32-025-30 HI172505015G10 HI17-10-010-5 HI17-18-036-20 HI17-25-050-30 HI17050052.5 HI17-25-010-60 HI17-53-150-60 HI17-20-100-10 HI17-25-025-60 HI17-20-040-40 HI17-18-040-20 HI17-53-010-15 HI17-32-010-60 HI17-53-150-75 HI17-10-040-5 HI17-32-010-15 HI1725100105 HI17-20-010-50 HI17-05-005-2 HI17-18-040-10 HI17-20-040-20 HI17-25-025-50 HI17-53-100-15 HI17-18-020-20 HI17-18-040-40 HI17-20-033-30 HI17-20-033-50 HI17-53-010-10 HI17-25-025-15 HI17-20-020-40 HI17-32-100-50 HI17-53-100-10 HI1732100105 HI17-20-040-10 HI172502560G5 HI172502530G5 HI17-10-020-10 HI17-15-015-5 HI17-20-020-60 HI17-10-010-10 HI17-10-020-15 HI17-25-050-50 HI17-25-025-30 HI17-25-050-10 HI17-15-015-15 HI17-32-150-50 HI17-53-050-60 HI17-53-025-15 HI17-18-020-60 HI17-20-020-10 HI17-25-025-10 HI172502515G5 HI17-25-100-50 HI17-32-100-30 HI17-53-150-50 HI17-20-010-40 HI17-20-100-60 HI17-53-150-10 HI17-15-015-20 HI17-15-015-10 HI172501030G5 HI17-32-150-30 HI17-32-100-60 HI17-20-100-50 HI172505030G10 HI17-25-100-30 HI17-20-010-10 HI17-18-040-60 HI17-18-020-10 HI17-10-040-15 HI17-20-010-20 HI17-32-010-50 HI17-53-100-50 HI17-53-025-50 HI17-32-050-30 HI17-32-025-60 HI17-18-020-40 HI17-53-100-60 HI17-32-025-15 HI17-20-100-20 HI17-53-050-30 HI17-20-033-25 HI17-18-020-50 HI17-53-050-15 HI172502530G10 HI17-25-100-60 HI17-53-025-30 HI17-32-100-15 HI17-18-018-60 HI1725050105 HI17-53-100-75 HI17-53-025-60 HI17-53-150-30
76432-690EA 1458.26 USD
76432-690 76432-692 76432-698 76432-852 76432-732 76432-854 76432-734 76432-694 76432-850 76432-696 76432-730 76432-728 76432-724 76432-846 76432-726 76432-848 76432-742 76432-864 76432-744 76432-866 76432-860 76432-740 76432-862 76432-856 76432-736 76432-858 76432-738 76432-790 76432-670 76433-998 76432-792 76432-830 76431-190 76432-676 76432-710 76432-798 76432-832 76432-678 76432-712 76432-672 76432-794 76432-674 76432-796 76432-706 76431-198 76432-828 76431-196 76432-708 76432-702 76431-194 76432-824 76432-704 76431-192 76432-826 76432-680 76432-682 76433-988 76432-720 76432-842 76432-688 76432-722 76432-844 76432-684 76432-686 76432-840 76432-838 76432-718 76432-834 76432-714 76432-836 76432-716 76433-992 76433-990 76433-996 76433-994 76433-978 76433-976 76432-770 76431-208 76432-654 76432-776 76432-656 76432-810 76432-778 76432-650 76432-772 76432-652 76432-774 76431-210 76432-806 76432-808 76432-768 76432-802 76432-648 76432-804 76433-982 76433-980 76433-986 76433-984 76432-780 76432-660 76432-786 76432-820 76432-666 76432-788 76432-700 76432-668 76432-822 76432-782 76432-662 76432-784 76432-664 76432-816 76434-008 76431-188 76432-818 76432-658 76432-812 76432-814 76432-874 76432-754 76434-000 76432-756 76434-002 76432-870 76432-750 76434-004 76432-872 76434-006 76432-752 76432-746 76432-868 76432-748 76432-764 76432-644 76432-766 76432-800 76432-646 76432-760 76432-640 76432-762 76432-642 76432-758 76432-638 76431-206 76431-204 76431-202 76431-200
Avantor® Hichrom HI-5 MS, GC Columns
Chromatography Columns
Really versatile general purpose low-polarity phase for a wide range of applications, including (for example): PCBs, essential oils, pesticides, semivolatiles and many more. HI-5 MS is an MS grade low-bleeding crossbonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: crossbonded 5% phenyl - 95% methyl polysiloxane - MS grade
  • Long lifetime
Max temperature limits may change depending on the film thickness.