Avantor® Hichrom HI-35 MS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI21-20-100-40 HI21-32-010-50 HI21-18-036-20 HI21-53-100-75 HI21-20-100-10 HI21-18-020-50 HI21-15-015-30 HI21-20-020-50 HI21-15-015-10 HI21-20-010-50 HI21-18-018-60 HI212505015G10 HI21-25-050-30 HI21-32-150-15 HI21-53-150-15 HI21-20-033-12 HI21-53-150-10 HI21-20-040-20 HI21-53-025-30 HI21-32-025-15 HI21-15-015-5 HI2132050105 HI21-25-050-15 HI21-32-025-30 HI212502560G10 HI21-18-020-20 HI21-10-020-15 HI21-25-010-30 HI21-25-100-50 HI21-32-010-15 HI21-20-010-60 HI21-10-020-10 HI21-53-100-15 HI21-20-010-40 HI21-32-100-15 HI212505030G5 HI21-32-050-15 HI2125025105 HI21-18-036-60 HI21-53-010-30 HI21-20-100-50 HI21-18-020-10 HI21-53-100-50 HI215310030G5 HI21-10-010-10 HI21050052.5 HI21-10-020-5 HI21-25-025-10 HI21-25-050-50 HI21-32-050-30 HI21-10-010-5 HI21-18-036-40 HI21-18-040-20 HI21-18-040-60 HI21-05-005-5 HI21-53-150-75 HI21-20-040-50 HI21-20-020-60 HI21-53-050-60 HI21-20-033-50 HI21-25-025-50 HI212502530G5 HI21-53-025-60 HI21-20-020-40 HI21-32-100-50 HI21-32-100-10 HI21-32-010-30 HI21-25-025-15 HI21-20-040-60 HI2132150105 HI21-18-020-40 HI2125100105 HI21-25-010-15 HI21-18-040-50 HI21-53-300-30 HI212505030G10 HI21-25-010-60 HI21-25-010-50 HI21-10-010-15 HI21-25-100-30 HI212501030G5 HI21-53-025-50 HI21-32-010-10 HI21-32-100-60 HI21-20-100-20 HI21-53-050-50 HI21-25-025-60 HI21-20-100-60 HI21-32-025-50 HI21-18-018-40 HI212502530G10 HI21-18-018-20 HI213202530G5 HI21-53-150-60 HI21-05-005-2 HI21-32-150-10 HI21-18-018-10 HI21-53-050-10 HI21-10-040-15 HI21-53-100-10 HI21-20-010-20 HI21-32-150-60 HI21-53-010-10 HI2132100105 HI21-32-010-60 HI21-32-025-10 HI21-20-020-10 HI21-53-050-15 HI2132010105 HI21-25-100-10 HI21-10-040-5 HI21-25-100-15 HI21-53-100-60 HI21-53-025-15 HI21-53-050-30 HI21-25-050-60 HI21-15-015-15 HI21-32-150-50 HI21-18-040-40 HI21-32-050-50 HI21-20-033-25 HI21-32-150-30 HI213210030G5 HI21-18-020-60 HI21-10-040-10 HI21-25-050-10 HI2125050105 HI21-20-033-30 HI21-53-010-15 HI21-18-018-50 HI21-32-050-60 HI21-20-040-40 HI21-15-015-20 HI21-32-025-60 HI21-32-050-10 HI21-53-100-30 HI21-53-150-30 HI212502515G5 HI21-20-040-10 HI21-20-010-10 HI2132025105 HI21-32-100-30 HI21-25-025-30 HI21-25-100-60 HI21-20-033-60 HI21-53-150-50 HI21-20-020-20 HI21-18-040-10 HI212502560G5
76432-570EA 898.76 USD
76432-570 76432-450 76432-572 76432-610 76432-456 76432-578 76432-612 76432-458 76432-452 76432-574 76432-454 76432-576 76432-608 76432-604 76432-606 76433-942 76432-580 76432-460 76432-582 76432-462 76432-500 76432-588 76432-622 76432-468 76432-502 76432-624 76432-584 76432-464 76432-586 76432-466 76432-620 76432-618 76432-614 76432-616 76431-148 76432-550 76432-434 76432-556 76432-436 76432-558 76432-430 76432-398 76432-552 76432-432 76432-554 76432-548 76432-428 76432-560 76432-440 76432-566 76432-600 76432-446 76432-568 76432-448 76432-602 76432-562 76432-442 76432-564 76432-444 76432-438 76432-492 76432-494 76432-490 76432-412 76432-534 76432-414 76432-536 76432-496 76432-530 76432-410 76432-498 76432-532 76432-526 76432-406 76431-170 76432-528 76432-408 76433-968 76433-966 76432-544 76432-424 76432-546 76432-426 76432-540 76432-420 76432-542 76432-422 76432-416 76432-538 76432-418 76433-970 76433-974 76433-972 76433-956 76432-470 76432-592 76433-954 76432-472 76432-594 76433-958 76432-590 76432-478 76432-632 76432-512 76432-634 76432-514 76432-474 76432-596 76432-476 76432-630 76432-510 76432-598 76432-508 76431-154 76431-152 76432-504 76431-150 76432-626 76432-506 76432-628 76433-960 76431-158 76433-964 76431-156 76433-962 76433-946 76432-482 76433-944 76432-484 76432-480 76433-948 76432-522 76432-402 76432-524 76432-404 76432-486 76432-520 76432-488 76432-400 76431-166 76431-164 76432-636 76431-162 76432-516 76431-160 76432-518 76433-952 76433-950 76431-168
Avantor® Hichrom HI-35 MS GC Columns
Chromatography Columns
Proprietary low-polarity and low-bleeding stationary phase; great choice for pesticides, herbicides, polychlorinated biphenyls (PCBs), and polyaromatic hydrocarbons (PAHs). Ideal for GC/MS applications and is also an excellent choice as a confirmation column for semivolatile environmental analysis.
  • Phase composition: Crossbonded low-polarity proprietary phase
  • Long lifetime
Max temperature limits may change depending on the film thickness.