Avantor® Hichrom HI-225 MS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI23-18-018-20 HI23050052.5 HI2332025105 HI23-25-025-50 HI23-20-020-40 HI23-53-025-60 HI23-53-025-30 HI23-05-005-5 HI23-32-025-30 HI23-20-010-40 HI23-10-010-15 HI23-20-010-60 HI2325025105 HI23-25-010-60 HI23-25-025-30 HI23-20-020-60 HI23-18-018-10 HI23-53-025-15 HI23-15-015-20 HI23-32-025-60 HI23-32-025-50 HI2332010105 HI23-25-010-15 HI23-18-018-60 HI23-20-020-10 HI23-32-010-10 HI23-32-010-30 HI23-32-025-10 HI23-15-015-15 HI23-20-010-10 HI23-32-010-60 HI23-53-010-10 HI23-53-010-15 HI23-10-010-10 HI23-25-025-60 HI23-18-018-50 HI23-05-005-2 HI23-18-018-40 HI23-10-010-5 HI23-32-010-15 HI23-53-025-50 HI23-32-010-50 HI23-25-025-15 HI23-20-020-50 HI23-20-010-20 HI23-15-015-10 HI23-20-020-20 HI23-20-010-50 HI23-53-010-30 HI23-32-025-15 HI23-15-015-30 HI23-25-010-50 HI23-15-015-5 HI23-25-025-10 HI23-25-010-30
76433-934EA 913.41 USD
76433-934 76432-372 76433-932 76432-374 76432-330 76433-938 76433-936 76432-370 76432-336 76432-338 76432-332 76432-376 76432-334 76432-378 76432-328 76433-940 76432-382 76433-924 76432-340 76432-384 76433-922 76433-928 76432-380 76433-926 76432-346 76432-348 76432-386 76432-342 76432-388 76432-344 76433-930 76432-350 76432-394 76432-396 76432-352 76432-390 76432-392 76432-358 76432-314 76432-316 76432-354 76432-310 76432-312 76432-356 76433-920 76432-360 76432-362 76432-324 76432-368 76432-326 76432-320 76432-364 76432-366 76432-322 76432-318
Avantor® Hichrom HI-225 MS GC Columns
Chromatography Columns
General purpose mid-to-high-polarity phase for a wide range of applications, like, for example: FAMEs, carbohydrates, sterols, flavor compounds. HI-225 MS is an MS grade crossbonded column.
  • Phase composition: Crossbonded 25% Cyanopropyl, 25% Phenyl, 50% Methyl Polysiloxane - MS
Max temperature limits may change depending on the film thickness.