Avantor® Hichrom HI-1 MS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI16-20-033-60 HI16-20-010-50 HI16-15-015-15 HI16-53-100-75 HI1625050105 HI16-10-040-10 HI16-53-010-30 HI16-18-036-60 HI16-25-025-15 HI16-32-100-10 HI16-20-033-12 HI16-18-020-10 HI16-20-020-10 HI16-25-100-50 HI162502515G5 HI16-32-100-50 HI16-32-025-15 HI16-32-010-60 HI16-53-150-50 HI16-20-020-40 HI16-32-010-50 HI16-10-020-15 HI1632010105 HI16-53-100-60 HI16-25-050-15 HI16-32-050-50 HI1632050105 HI16-32-050-30 HI16-25-100-10 HI16-10-040-15 HI16-20-040-40 HI16-18-020-40 HI16-53-025-15 HI16-53-150-75 HI1625100105 HI16-18-040-10 HI16-10-020-5 HI16-32-150-50 HI16-53-150-10 HI16-32-025-10 HI16-18-040-60 HI16-32-025-50 HI16-20-100-10 HI16-32-100-60 HI16-53-025-60 HI16-18-036-20 HI16-32-025-30 HI1625025105 HI16-53-100-15 HI16-18-018-20 HI16-32-010-30 HI16-32-150-30 HI16-20-020-20 HI16-20-100-60 HI16-05-005-2 HI16-25-100-15 HI16-32-100-30 HI16-32-050-60 HI16-32-150-10 HI16-20-010-40 HI16-53-050-30 HI16-20-040-10 HI16-18-018-10 HI16-10-010-15 HI16-20-040-20 HI16-15-015-5 HI16-18-020-50 HI16-20-100-50 HI16-05-005-5 HI1632150105 HI16-10-010-5 HI16-25-050-30 HI16-18-018-40 HI16-53-025-30 HI16-20-020-50 HI162502560G5 HI16-32-100-15 HI165310030G5 HI16-53-050-10 HI16050052.5 HI16-10-010-10 HI16-18-040-50 HI16-53-050-50 HI16-18-040-20 HI16-25-025-30 HI16-25-025-50 HI16-53-150-15 HI16-32-150-60 HI16-25-010-30 HI16-18-020-20 HI16-20-033-25 HI16-25-010-60 HI16-18-020-60 HI16-18-040-40 HI16-25-050-60 HI16-20-033-30 HI16-32-150-15 HI16-53-010-10 HI16-32-010-10 HI16-10-040-5 HI16-25-100-30 HI16-20-100-40 HI16-53-100-50 HI16-18-018-60 HI16-18-036-40 HI16-20-033-50 HI16-32-050-10 HI16-25-025-60 HI16-53-150-30 HI16-53-100-10 HI16-20-040-60 HI16-15-015-10 HI1632025105 HI16-25-010-15 HI16-25-100-60 HI16-53-025-50 HI16-53-100-30 HI16-20-010-10 HI16-20-010-60 HI16-18-018-50 HI16-15-015-30 HI16-25-025-10 HI162502530G5 HI16-25-050-10 HI16-53-150-60 HI16-53-050-60 HI16-20-020-60 HI16-25-010-50 HI16-32-010-15 HI16-15-015-20 HI16-32-050-15 HI1632100105 HI16-20-040-50 HI16-20-100-20 HI16-25-050-50 HI16-20-010-20 HI16-53-010-15 HI16-32-025-60 HI16-53-050-15 HI16-10-020-10
76432-052EA 1082.25 USD
76432-052 76432-054 76431-928 76431-926 76432-050 76431-924 76432-056 76432-058 76431-892 76431-130 76431-890 76431-932 76431-930 76431-898 76431-896 76431-894 76432-064 76431-938 76432-066 76431-936 76432-060 76431-934 76432-062 76432-068 76431-944 76431-942 76431-940 76432-030 76431-908 76432-032 76433-878 76431-906 76431-904 76431-868 76431-902 76432-038 76432-034 76432-036 76431-990 76431-870 76431-910 76431-998 76433-882 76431-878 76431-996 76433-880 76431-876 76431-994 76431-874 76433-884 76431-992 76431-872 76433-868 76431-918 76432-042 76433-866 76431-916 76432-044 76431-914 76431-912 76432-040 76432-046 76432-048 76431-880 76431-888 76433-872 76431-922 76431-886 76433-870 76431-128 76431-920 76431-884 76433-876 76431-126 76431-882 76431-124 76433-874 76433-858 76432-010 76433-856 76431-848 76431-846 76431-968 76432-016 76432-018 76432-012 76432-014 76431-976 76433-860 76431-856 76431-974 76431-854 76431-972 76433-864 76431-852 76431-970 76433-862 76431-850 76432-020 76432-022 76431-978 76431-858 76432-028 76432-024 76432-026 76431-980 76431-866 76431-900 76431-988 76431-864 76431-986 76431-862 76431-984 76433-854 76431-860 76431-982 76433-852 76432-070 76431-948 76431-946 76431-954 76431-834 76431-952 76431-950 76432-000 76431-958 76431-838 76431-956 76431-836 76432-006 76432-008 76432-002 76432-004 76431-844 76431-966 76431-842 76431-964 76431-840 76431-962 76431-960
Avantor® Hichrom HI-1 MS GC Columns
Chromatography Columns
Really versatile general purpose apolar phase for a wide range of MS applications, including (for example): PCBs, simulated distillation, gases, essential oils, hydrocarbons analysis, pesticides, semi volatiles, oxygenates. HI-1 MS is an MS grade low-bleeding cross bonded column and assure a long lifetime even under high temperature limits.
  • Phase composition: Cross bonded 100% methyl polysiloxane (100% dimethylpolysiloxane) - MS grade
  • Long lifetime
  • Equivalent to USP phases G1, G2, G38
Max temperature limits may change depending on the film thickness.