Avantor® Hichrom HI-SE54 HT GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI91-53-010-15 HI91-32-015-30 HI91-32-025-30 HI91-53-025-30 HI91-10-010-10 HI91-53-010-30 HI91-32-025-15 HI91-53-025-15 HI91-32-010-10 HI91-25-010-15 HI91-10-010-5 HI91-25-025-15 HI91-32-010-30 HI91-32-015-15 HI91-32-010-15 HI91-53-017-30 HI91-25-010-30 HI91-53-010-10 HI91-25-015-15 HI91-53-015-15 HI91-25-025-30 HI91-25-015-30 HI91-53-015-30 HI913201010G2
76431-746EA 768.04 USD
76431-746 76431-738 76431-742 76431-758 76431-826 76431-748 76431-740 76431-756 76431-730 76431-718 76431-828 76431-726 76431-734 76431-736 76431-732 76431-754 76431-720 76431-744 76431-722 76431-750 76431-728 76431-724 76431-752 76431-422
Avantor® Hichrom HI-SE54 HT GC Columns
Chromatography Columns
High Temperature general purpose column for high boiling petroleum products, Long-chained hydrocarbons.
Max temperature limits may change depending on the film thickness.