Avantor® Hichrom HI-8 HT GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI88-10-010-10 HI88-25-015-15 HI883201010G2 HI88-25-010-15 HI88-25-025-15 HI88-10-010-5 HI88-32-015-15 HI88-25-015-30 HI88-32-025-15 HI88-53-010-30 HI88-53-017-30 HI88-53-010-10 HI88-32-010-30 HI88-25-010-30 HI88-53-015-30 HI88-53-025-15 HI88-53-010-15 HI88-32-010-15 HI88-32-015-30 HI88-32-010-10 HI88-25-025-30 HI88-32-025-30 HI88-53-025-30 HI88-53-015-15
76431-822EA 1268.79 USD
76431-822 76431-680 76431-292 76431-676 76431-684 76431-824 76431-694 76431-682 76431-698 76431-706 76431-712 76431-702 76431-692 76431-678 76431-710 76431-714 76431-704 76431-690 76431-696 76431-688 76431-686 76431-700 76431-716 76431-708
Avantor® Hichrom HI-8 HT GC Columns
Chromatography Columns
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Maximum temperature limits may change depending on the film thickness.