Avantor® Hichrom HI-1701 HT, GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI92-53-010-30 HI92-10-010-10 HI92-10-010-5 HI92-53-015-15 HI92-25-010-30 HI92-32-010-15 HI92-25-025-15 HI92-25-025-30 HI92-32-025-15 HI92-32-025-30 HI92-53-017-30 HI92-25-010-15 HI92-32-010-10 HI92-53-010-15 HI92-53-025-15 HI92-32-015-15 HI92-53-015-30 HI92-32-010-30 HI92-53-025-30 HI92-25-015-15 HI92-25-015-30 HI92-32-015-30 HI92-53-010-10
76431-570EA 1142.5 USD
76431-570 76431-810 76431-812 76431-572 76431-542 76431-554 76431-548 76431-550 76431-562 76431-564 76431-576 76431-540 76431-552 76431-568 76431-578 76431-558 76431-574 76431-556 76431-580 76431-544 76431-546 76431-560 76431-566
Avantor® Hichrom HI-1701 HT, GC Columns
Chromatography Columns
High temperature general purpose column. Ideal for confrmation analysis, ideal as mid-polar column in GC×GC-HT confgurations.
Max temperature limits may change depending on the film thickness.