Avantor® Hichrom HI-PLUS GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI50-25-025-50 HI50-25-025-60 HI50-25-100-10 HI50-20-010-20 HI50-05-005-5 HI50-15-015-30 HI50-18-018-60 HI50-32-100-50 HI50-18-018-40 HI50-32-050-60 HI50-53-050-60 HI50-25-050-60 HI50-18-040-40 HI50-32-050-30 HI50-32-050-10 HI50-53-100-60 HI50-53-150-50 HI50-53-150-10 HI50-32-010-50 HI50-53-100-50 HI5025025105 HI50-25-010-50 HI502502530G5 HI50-53-050-30 HI50-53-025-50 HI5032050105 HI50-32-025-10 HI50-32-010-30 HI50-53-150-60 HI50-10-040-15 HI50-20-040-20 HI50-20-040-60 HI50-32-025-30 HI50-18-020-10 HI50-10-020-10 HI50-53-100-30 HI50-32-025-50 HI50-32-010-10 HI50-25-050-50 HI50-53-100-15 HI50-32-050-15 HI50-32-100-30 HI50-20-040-10 HI50-10-020-15 HI50-25-010-60 HI50-18-018-20 HI50-20-100-60 HI5032010105 HI50-53-025-15 HI50-18-020-40 HI5032100105 HI50-25-025-10 HI50-05-005-2 HI50-10-010-15 HI50050052.5 HI50-25-025-15 HI50-32-100-60 HI50-32-100-15 HI50-32-025-15 HI50-32-010-15 HI50-20-100-50 HI50-20-020-50 HI502502515G5 HI50-25-025-30 HI50-53-050-50 HI50-53-025-60 HI50-10-040-10 HI50-25-050-10 HI50-53-050-10 HI50-10-020-5 HI50-20-040-50 HI50-20-100-10 HI50-25-050-30 HI50-53-150-75 HI50-20-010-40 HI50-32-010-60 HI50-20-100-40 HI50-18-040-50 HI50-32-025-60 HI50-15-015-10 HI50-53-050-15 HI50-20-010-10 HI50-53-010-15 HI50-10-010-10 HI50-25-050-15 HI50-53-025-30 HI50-20-040-40 HI50-32-100-10 HI50-20-010-50 HI50-53-150-15 HI50-25-100-60 HI50-53-100-75 HI50-53-100-10 HI5025050105 HI50-25-010-30 HI50-25-010-15 HI50-20-020-20 HI50-18-018-10 HI50-15-015-20 HI50-53-010-10 HI50-20-020-10 HI50-10-040-5 HI50-20-020-40 HI5025100105 HI505310030G5 HI50-10-010-5 HI50-15-015-15 HI50-20-010-60 HI50-32-050-50 HI50-25-100-15 HI50-20-020-60 HI50-25-100-30 HI50-18-040-10 HI50-18-018-50 HI50-25-100-50 HI50-18-040-20 HI50-20-100-20 HI50-53-010-30 HI50-15-015-5 HI50-53-150-30 HI5032025105 HI50-18-020-20 HI50-18-040-60
76427-304EA 1114.57 USD
76427-304 76427-306 76427-268 76427-146 76427-300 76427-302 76427-148 76427-308 76427-154 76427-274 76427-310 76427-156 76427-276 76427-150 76427-270 76427-152 76427-272 76427-256 76427-258 76427-264 76427-142 76427-266 76427-144 76427-260 76427-262 76430-620 76430-622 76427-246 76430-624 76427-248 76430-626 76430-616 76427-252 76430-618 76427-254 76427-250 76427-238 76430-610 76430-612 76427-234 76430-614 76427-236 76427-242 76427-244 76430-608 76427-240 76427-228 76427-224 76427-344 76427-226 76427-346 76430-640 76427-190 76427-192 76430-638 76427-198 76427-230 76427-232 76427-194 76427-196 76427-338 76430-630 76427-216 76430-632 76427-218 76427-334 76430-634 76427-212 76427-336 76430-636 76427-214 76427-180 76427-220 76430-628 76427-186 76427-340 76427-222 76427-342 76427-188 76427-182 76427-184 76427-206 76427-326 76427-208 76427-328 76427-202 76427-322 76427-168 76427-204 76427-324 76427-170 76427-290 76427-330 76427-176 76427-296 76427-332 76427-178 76427-298 76427-210 76427-172 76427-292 76427-174 76427-294 76427-316 76427-318 76427-312 76427-158 76427-278 76431-318 76427-314 76431-320 76427-280 76427-286 76427-164 76427-288 76427-200 76427-320 76427-166 76427-282 76431-324 76427-160 76427-284 76427-162
Avantor® Hichrom HI-PLUS GC Columns
Chromatography Columns
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Maximum temperature limits may change depending on the film thickness.