Avantor® Hichrom HI-DEX G-01 GC columns
Supplier: Avantor®
Avantor® Hichrom
HI74-25-015-25 HI74-25-015-15 HI74-25-025-15 HI84-10-010-5 HI74-25-015-60 HI74-25-025-25 HI74-25-025-30 HI74-25-025-60 HI74-25-015-50 HI74-25-015-30 HI74-25-025-50 HI84-10-010-10
76430-014EA 1527.24 USD
76430-014 76430-026 76430-016 76430-028 76430-018 76430-030 76430-020 76430-086 76430-032 76430-088 76430-022 76430-024
Avantor® Hichrom HI-DEX G-01 GC columns
Chromatography Columns
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.