Avantor® Hichrom HI-DEX DMT Beta GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI71-25-015-15 HI83-10-010-5 HI71-25-025-50 HI71-25-015-50 HI83-10-010-10 HI71-25-025-15 HI71-25-025-25 HI71-25-015-60 HI71-25-015-30 HI71-25-025-30 HI71-25-025-60 HI71-25-015-25
76429-994EA 1089.63 USD
76429-994 76430-084 76430-010 76430-000 76430-082 76430-004 76430-006 76430-002 76429-998 76430-008 76430-012 76429-996
Avantor® Hichrom HI-DEX DMT Beta GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.