Avantor® Hichrom HI-DEX DET Beta GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI80-10-010-5 HI68-25-015-60 HI68-25-015-25 HI68-25-025-60 HI68-25-015-30 HI68-25-025-30 HI68-25-025-25 HI80-10-010-10 HI68-25-025-50 HI68-25-025-15 HI68-25-015-15 HI68-25-015-50
76430-072EA 1089.63 USD
76430-072 76429-942 76429-936 76429-952 76429-938 76429-948 76429-946 76430-070 76429-950 76429-944 76429-934 76429-940
Avantor® Hichrom HI-DEX DET Beta GC Columns
Chromatography Columns
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.