Avantor® Hichrom HI-PS255, GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI58-15-015-20 HI58-32-100-60 HI58-20-100-60 HI58-53-300-15 HI58-32-050-50 HI58-53-500-30 HI58-10-010-15 HI58-53-100-30 HI58-32-400-30 HI58-32-300-30 HI58-32-025-60 HI58-53-300-75 HI58-20-010-20 HI58-20-100-50 HI58-25-100-15 HI58-15-015-10 HI58-32-010-50 HI58-20-010-60 HI58-18-018-20 HI58-20-020-10 HI58-32-010-10 HI58-32-500-60 HI58-25-300-60 HI58150043G2 HI58-18-018-10 HI58-53-010-15 HI58-53-050-30 HI58-53-050-60 HI58-20-020-40 HI58-53-300-10 HI5832300105 HI58-20-010-10 HI58-32-100-30 HI5832100105 HI58-25-050-50 HI5832010105 HI58-20-040-50 HI58-25-300-10 HI58-25-100-30 HI5825050105 HI58-25-010-15 HI58-53-500-50 HI58-20-100-20 HI58-25-050-60 HI58-53-100-50 HI58-53-150-30 HI58-25-200-15 HI58-20-010-40 HI58-53-100-60 HI585310030G5 HI58-53-150-60 HI58-32-300-60 HI58-32-100-15 HI58-32-025-10 HI58-32-150-50 HI58-18-018-50 HI58-25-025-30 HI58-20-020-60 HI58-53-500-10 HI58-53-025-30 HI58-20-040-10 HI58-25-010-60 HI58-53-025-60 HI58-20-100-10 HI58-25-025-10 HI58-25-300-15 HI58-25-300-30 HI58-25-025-15 HI58-32-100-10 HI58-32-500-30 HI58-18-040-20 HI58-53-300-60 HI58-10-010-10 HI58-53-300-50 HI58-25-100-50 HI5825025105 HI58-32-150-30 HI58-15-015-5 HI58-32-050-30 HI58-32-025-30 HI58-10-010-5 HI58-18-020-20 HI58-53-050-50 HI58-10-040-10 HI58-53-700-15 HI58-32-010-15 HI58-10-040-5 HI58-25-200-50 HI58-53-700-30 HI5832050105 HI58-53-010-10 HI58050052.5 HI58-32-025-15 HI58-15-015-30 HI58-32-300-50 HI58-53-300-30 HI58-32-010-60 HI58-32-010-30 HI58-18-040-50 HI58-53-010-30 HI58-20-040-40 HI58-05-005-2 HI582502515G5 HI58-18-040-40 HI58-10-040-15 HI58-18-040-60 HI58-25-200-60 HI58-53-500-75 HI58-53-050-10 HI58-20-010-50 HI58-18-040-10 HI58-32-500-50 HI58-25-100-10 HI58-53-265-10 HI58-53-150-75 HI58-25-010-30 HI58-53-025-15 HI58-25-025-60 HI58-20-040-60 HI58-32-150-60 HI58-53-050-15 HI58-53-025-50 HI58-32-025-50 HI58-20-020-50 HI58-10-020-15 HI582502560G5 HI5832500105 HI58-18-020-40 HI58-20-020-20 HI58-53-500-60 HI5825200105 HI58-32-050-60 HI58-53-100-15 HI582502530G5 HI58-32-500-15 HI58-32-050-10 HI58-25-025-50 HI58-25-200-30 HI58-25-010-50 HI58-32-150-15 HI58-25-050-10 HI58-53-150-15 HI58-20-100-40 HI5825100105 HI58-15-004-10 HI58-25-200-10 HI58-32-150-10 HI58-53-150-10 HI58-53-500-15 HI5832150105 HI58-25-050-15 HI58-32-500-10 HI58-32-300-15 HI58-32-050-15 HI58-20-040-20 HI58-15-015-15 HI58-10-020-10 HI58-10-020-5 HI5832025105 HI58-25-300-50 HI58-05-005-5 HI58-18-018-40 HI58-53-150-50 HI58-53-100-75 HI58-18-018-60 HI58-32-300-10 HI58-32-100-50 HI58-25-050-30 HI58-25-100-60 HI58-53-100-10 HI58-18-020-10
76430-806EA 1129.27 USD
76430-806 76428-404 76428-290 76428-496 76428-390 76428-510 76430-786 76428-472 76428-430 76428-424 76428-380 76428-504 76428-254 76428-288 76428-328 76430-802 76428-366 76428-260 76428-228 76428-262 76428-358 76428-442 76428-356 76431-358 76428-226 76428-446 76428-462 76428-466 76428-266 76428-494 76428-420 76428-252 76428-400 76428-396 76428-320 76428-360 76428-278 76428-348 76428-330 76428-314 76428-292 76428-512 76428-284 76428-322 76428-474 76428-484 76428-340 76428-256 76428-476 76431-366 76428-488 76428-428 76428-398 76428-370 76428-414 76428-232 76428-306 76428-270 76428-506 76428-452 76428-272 76428-298 76428-456 76428-282 76428-300 76428-350 76428-352 76428-304 76428-394 76428-438 76428-244 76428-502 76430-784 76428-500 76428-332 76428-302 76428-412 76430-810 76428-388 76428-376 76430-788 76428-238 76428-464 76430-796 76428-518 76428-362 76430-800 76428-344 76428-520 76428-384 76428-444 76430-780 76428-374 76430-808 76428-426 76428-498 76428-368 76428-364 76428-248 76428-448 76428-276 76430-778 76431-360 76428-246 76430-798 76428-250 76428-346 76428-516 76428-458 76428-258 76428-242 76428-440 76428-324 76428-492 76428-490 76428-294 76428-450 76428-310 76428-280 76428-416 76428-460 76428-454 76428-378 76428-268 76430-792 76431-364 76428-434 76428-240 76428-264 76428-514 76428-338 76428-392 76428-470 76431-362 76428-436 76428-382 76428-308 76428-342 76428-296 76428-410 76428-312 76428-482 76428-286 76428-326 76428-224 76428-336 76428-406 76428-480 76428-508 76428-408 76428-316 76428-432 76428-422 76428-386 76428-274 76430-804 76430-790 76430-794 76428-372 76428-354 76430-782 76428-230 76428-486 76428-478 76428-234 76428-418 76428-402 76428-318 76428-334 76428-468 76428-236
Avantor® Hichrom HI-PS255, GC Columns
Chromatography Columns
Apolar phase, suitable for high flm thickness columns, to analyze solvents, alcohols, volatiles, and so on.
Max temperature limits may change depending on the film thickness.