Avantor® Hichrom HI-POF 1 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI55-25-015-60 HI55-18-018-20 HI55-25-025-30 HI55-32-050-30 HI55-32-025-30 HI55-25-015-30 HI55-25-025-15 HI55-32-032-30 HI55-53-050-30 HI55-25-025-60 HI55-32-050-15
76428-190EA 1304.05 USD
76428-190 76428-180 76428-178 76428-174 76428-186 76428-176 76428-188 76428-192 76428-182 76428-172 76428-184
Avantor® Hichrom HI-POF 1 GC Columns
Chromatography Columns
Application specifc columns developed for pesticides, herbicides, insecticides analysis and so on.
Max temperature limits may change depending on the film thickness.