Avantor® Hichrom HI-PLUS 75 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI5332100105 HI53-18-018-60 HI53-18-040-10 HI53-53-010-10 HI53-25-025-15 HI53-32-100-10 HI53-20-010-40 HI53-53-010-30 HI53-53-100-75 HI5325025105 HI53-15-015-10 HI53-25-010-30 HI53-15-015-5 HI53-25-025-10 HI53-20-020-60 HI53-20-040-50 HI53-53-150-10 HI53-10-010-5 HI53-15-015-20 HI5332010105 HI53-10-020-15 HI53-10-020-5 HI53-25-010-15 HI53050052.5 HI53-32-100-30 HI53-18-040-40 HI532502515G5 HI53-32-050-30 HI53-18-018-20 HI53-32-010-10 HI53-25-025-50 HI53-32-025-60 HI53-20-010-60 HI53-20-100-40 HI532502530G5 HI53-25-100-30 HI53-18-040-50 HI53-32-100-50 HI53-53-025-30 HI53-53-025-60 HI53-18-040-60 HI53-18-018-10 HI53-53-010-15 HI53-25-100-10 HI53-20-020-10 HI53-20-040-40 HI53-20-020-20 HI53-53-100-50 HI53-18-018-40 HI53-25-050-15 HI53-15-015-30 HI53-53-150-50 HI53-25-025-60 HI53-53-025-15 HI53-32-025-15 HI53-25-010-50 HI53-53-100-15 HI53-32-010-60 HI53-32-100-60 HI53-25-050-30 HI53-20-040-10 HI53-32-025-30 HI53-10-040-15 HI53-05-005-5 HI53-53-100-60 HI53-32-050-15 HI532502560G5 HI53-18-020-20 HI53-10-010-10 HI5332025105 HI53-53-150-75 HI53-32-100-15 HI53-20-100-50 HI53-32-025-50 HI53-10-010-15 HI53-53-150-30 HI53-53-050-15 HI53-20-100-60 HI53-32-050-10 HI53-32-010-30 HI53-53-100-10 HI53-20-010-20 HI53-53-025-50 HI53-53-050-60 HI53-32-050-50 HI53-53-050-10 HI53-20-020-50 HI535310030G5 HI53-20-100-10 HI53-32-010-50 HI53-20-020-40 HI53-18-018-50 HI5325100105 HI53-18-020-40 HI53-25-100-50 HI53-32-025-10 HI53-25-100-60 HI53-32-010-15 HI53-18-020-10 HI53-53-150-60 HI53-25-010-60 HI53-20-010-50 HI53-25-100-15 HI53-20-100-20 HI53-10-020-10 HI53-25-050-10 HI53-32-050-60 HI53-53-150-15 HI5332050105 HI53-20-040-60 HI53-15-015-15 HI53-05-005-2 HI53-18-040-20 HI53-53-100-30 HI53-53-050-30 HI53-25-050-60 HI53-20-040-20 HI53-25-050-50 HI53-20-010-10 HI53-53-050-50 HI53-25-025-30 HI53-10-040-10 HI5325050105 HI53-10-040-5
76427-944EA 748.96 USD
76427-944 76427-824 76427-788 76430-720 76427-942 76427-822 76427-940 76430-722 76427-820 76427-784 76430-724 76427-786 76427-948 76427-828 76427-946 76427-826 76427-792 76430-716 76427-794 76430-718 76427-790 76427-812 76427-778 76427-934 76427-810 76427-898 76427-932 76430-710 76427-774 76427-896 76430-712 76427-930 76427-776 76427-894 76430-714 76427-818 76427-816 76427-938 76427-814 76427-936 76427-780 76427-782 76427-922 76430-740 76427-802 76427-766 76430-742 76427-920 76427-768 76427-888 76427-800 76427-762 76427-886 76427-764 76427-884 76427-928 76427-808 76427-926 76427-806 76427-924 76427-804 76427-892 76427-770 76430-738 76427-772 76427-890 76427-878 76427-912 76430-730 76427-876 76427-910 76430-732 76427-874 76430-734 76427-872 76430-736 76427-918 76427-916 76427-914 76430-726 76427-882 76430-728 76427-760 76427-880 76427-900 76427-868 76427-866 76427-864 76427-862 76427-908 76427-906 76427-904 76427-902 76427-870 76427-856 76427-854 76427-852 76427-850 76427-858 76431-342 76427-860 76431-348 76431-346 76431-344 76427-846 76427-964 76427-844 76427-962 76427-842 76427-960 76427-840 76427-848 76427-834 76427-956 76427-832 76427-954 76427-796 76427-830 76427-952 76427-798 76427-950 76427-838 76427-836 76427-958
Avantor® Hichrom HI-PLUS 75 GC Columns
Chromatography Columns
Offering a range of selectivities with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Max temperature limits may change depending on the film thickness.