Avantor® Hichrom HI-PLUS 25 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI52-32-100-15 HI52-53-100-15 HI52-15-015-30 HI52-53-050-50 HI52-53-025-15 HI52-20-020-40 HI52050052.5 HI52-32-100-30 HI52-25-100-50 HI522502515G5 HI525310030G5 HI52-18-018-10 HI52-10-010-10 HI52-32-010-15 HI52-32-050-50 HI52-32-010-30 HI52-18-018-40 HI52-25-025-15 HI522502560G5 HI52-25-100-60 HI52-53-050-60 HI52-32-025-15 HI52-53-150-50 HI52-20-040-50 HI52-20-010-10 HI52-53-150-30 HI52-15-015-10 HI52-53-100-60 HI52-18-018-50 HI5232025105 HI52-53-010-15 HI52-32-010-10 HI52-53-050-10 HI52-18-018-20 HI52-32-010-60 HI52-53-100-30 HI52-53-150-10 HI52-53-025-50 HI52-18-040-60 HI52-32-025-30 HI52-32-100-50 HI52-20-100-40 HI52-18-040-50 HI52-53-150-15 HI52-18-040-10 HI52-20-020-10 HI52-20-010-40 HI52-25-050-30 HI52-18-020-10 HI52-25-100-15 HI52-53-150-75 HI52-32-025-60 HI52-25-025-30 HI52-10-010-5 HI5232010105 HI52-53-100-50 HI52-25-100-30 HI52-20-040-10 HI52-25-050-10 HI52-25-050-60 HI52-53-050-15 HI52-53-100-10 HI52-32-050-15 HI52-05-005-5 HI5232100105 HI5225025105 HI52-25-010-60 HI52-18-020-20 HI52-15-015-5 HI52-20-040-40 HI52-53-150-60 HI52-18-040-20 HI52-53-100-75 HI52-15-015-15 HI52-20-040-20 HI52-53-010-30 HI52-10-040-5 HI52-20-010-60 HI52-25-010-30 HI52-10-020-15 HI52-32-010-50 HI52-10-040-10 HI52-25-100-10 HI52-32-100-10 HI52-53-050-30 HI52-25-050-15 HI52-20-020-50 HI522502530G5 HI52-32-050-60 HI52-25-010-15 HI52-53-025-30 HI52-20-020-20 HI52-25-025-60 HI52-25-025-10 HI52-53-010-10 HI52-18-040-40 HI52-20-100-60 HI52-20-010-20 HI52-25-050-50 HI52-32-050-30 HI52-32-025-10 HI52-20-100-20 HI52-10-010-15 HI52-32-100-60 HI52-20-100-50 HI52-20-010-50 HI52-10-020-10 HI52-18-018-60 HI52-10-040-15 HI52-15-015-20 HI52-20-100-10 HI52-32-025-50 HI52-25-010-50 HI5225050105 HI52-10-020-5 HI52-25-025-50 HI52-20-040-60 HI5225100105 HI5232050105 HI52-53-025-60 HI52-05-005-2 HI52-20-020-60 HI52-32-050-10 HI52-18-020-40
76427-668EA 701.96 USD
76427-668 76430-686 76427-700 76427-702 76430-688 76427-664 76427-666 76427-708 76430-680 76427-704 76430-682 76427-706 76430-684 76427-670 76427-672 76427-656 76430-676 76427-658 76430-678 76427-652 76427-654 76427-660 76430-704 76430-706 76427-662 76430-708 76427-646 76427-648 76427-642 76427-644 76427-650 76427-756 76430-696 76427-634 76427-758 76430-698 76427-636 76427-752 76427-598 76427-630 76427-754 76427-632 76430-690 76430-692 76427-638 76430-694 76427-640 76427-624 76427-744 76427-626 76427-746 76427-620 76427-740 76427-586 76427-622 76427-742 76427-588 76427-628 76427-748 76427-594 76431-338 76427-750 76427-596 76431-336 76431-334 76427-590 76427-592 76427-734 76427-612 76427-736 76427-614 76427-576 76427-730 76427-696 76427-732 76427-578 76427-698 76427-610 76427-738 76427-616 76427-618 76431-340 76427-582 76427-584 76427-580 76427-602 76427-722 76427-568 76427-604 76427-724 76430-700 76427-686 76427-564 76427-600 76430-702 76427-688 76427-566 76427-720 76427-606 76427-726 76427-608 76427-728 76427-572 76427-692 76427-574 76427-694 76427-570 76427-690 76427-558 76427-712 76427-678 76427-714 76427-554 76427-674 76427-556 76427-710 76427-676 76427-716 76427-718 76427-682 76427-560 76427-684 76427-562 76427-680
Avantor® Hichrom HI-PLUS 25 GC Columns
Chromatography Columns
Offering a range of selectivity's with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Max temperature limits may change depending on the film thickness.