Avantor® Hichrom HI-FFAP EXT GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI44-53-200-60 HI44-05-005-2 HI44-20-100-50 HI44-53-200-15 HI44-20-040-50 HI44-32-010-15 HI44-53-100-30 HI44-18-040-10 HI44-25-025-60 HI44-20-100-10 HI442502515G5 HI44-10-010-5 HI44-53-025-60 HI44-53-250-15 HI44-20-040-10 HI44-32-150-30 HI44-25-025-15 HI44-53-025-50 HI44-25-010-30 HI44-18-018-50 HI44-20-020-60 HI44-53-250-50 HI4432050105 HI44-53-010-30 HI44-10-010-10 HI44-53-150-30 HI44-53-150-50 HI44-32-050-30 HI44-32-025-15 HI44-25-050-60 HI44-18-018-20 HI44-10-020-5 HI44-25-100-50 HI44-20-010-40 HI44-32-150-50 HI44-25-025-10 HI44-32-100-60 HI44-53-250-60 HI44-18-040-50 HI44-32-010-50 HI44-25-025-30 HI44-25-100-60 HI44-32-025-50 HI4432100105 HI44-10-040-10 HI4432150105 HI44-18-040-40 HI44-32-010-10 HI44-32-050-15 HI44-53-050-50 HI44-53-250-75 HI44-32-300-60 HI44-18-020-60 HI44-20-040-25 HI44-53-250-30 HI44-32-025-60 HI44-53-100-75 HI44-20-020-50 HI44-32-025-30 HI44050052.5 HI4432300105 HI4425100105 HI44-32-050-60 HI445310030G5 HI4432010105 HI44-25-100-10 HI44-32-150-60 HI44-32-100-15 HI442502530G5 HI44-53-150-15 HI44-32-300-50 HI44-20-040-40 HI44-32-150-15 HI44-53-200-50 HI4425025105 HI44-20-010-60 HI44-53-050-15 HI44-32-050-10 HI44-25-025-50 HI44-32-025-10 HI44-15-015-15 HI44-25-100-15 HI44-32-150-10 HI44-25-100-30 HI44-15-015-30 HI44-20-020-40 HI44-20-010-10 HI44-18-018-10 HI44-25-010-15 HI44-18-020-10 HI44-32-050-50 HI44-20-100-20 HI442502560G5 HI44-53-100-15 HI44-32-100-50 HI44-53-200-30 HI44-20-040-20 HI44-10-040-5 HI44-25-050-50 HI44-53-050-10 HI44-18-040-20 HI44-53-150-10 HI44-32-010-30 HI44-53-025-15 HI44-20-100-60 HI44-53-100-50 HI44-53-100-10 HI44-32-300-15 HI44-53-025-30 HI44-53-100-60 HI44-20-010-50 HI44-32-300-30 HI44-32-100-10 HI44-20-040-60 HI44-20-020-10 HI44-53-200-75 HI44-32-300-10 HI44-20-020-20 HI44-10-020-10 HI44-18-018-40 HI44-25-050-30 HI44-05-005-5 HI44-18-020-20 HI44-53-050-30 HI44-18-020-50 HI44-15-015-20 HI44-15-015-5 HI44-10-020-15 HI4425050105 HI44-53-250-10 HI44-10-040-15 HI44-18-020-40 HI44-18-040-60 HI44-53-050-60 HI44-15-015-10 HI44-53-200-10 HI44-25-050-15 HI44-25-010-60 HI44-32-010-60 HI44-53-150-60 HI44-53-150-75 HI44-20-010-20 HI44-53-010-15 HI4432025105 HI44-20-100-40 HI44-18-018-60 HI44-25-010-50 HI44-53-010-10 HI44-25-050-10 HI44-32-100-30 HI44-10-010-15
76430-564EA 791.51 USD
76430-564 76430-566 76430-568 76426-740 76426-586 76430-560 76430-562 76426-616 76426-736 76426-614 76426-734 76426-612 76426-732 76426-698 76426-610 76426-730 76426-618 76426-738 76430-554 76430-556 76430-558 76426-630 76426-750 76426-596 76426-594 76430-550 76426-592 76430-552 76426-590 76426-748 76426-626 76426-746 76426-624 76426-744 76426-622 76426-588 76426-742 76426-620 76431-306 76431-304 76431-302 76426-628 76426-684 76426-682 76426-680 76426-836 76426-714 76426-834 76426-712 76426-832 76426-678 76426-710 76426-798 76426-830 76426-676 76426-796 76426-718 76426-838 76426-716 76426-696 76426-694 76430-570 76426-692 76430-572 76426-690 76426-726 76426-604 76426-724 76426-844 76426-602 76426-722 76426-688 76426-842 76426-600 76426-720 76426-840 76426-686 76426-608 76426-728 76426-606 76426-784 76426-662 76426-782 76426-660 76426-780 76426-814 76426-812 76426-658 76426-778 76426-810 76426-656 76426-776 76426-654 76426-774 76426-818 76426-816 76426-674 76426-794 76426-672 76426-792 76426-670 76426-790 76426-704 76426-824 76426-702 76426-668 76426-822 76426-700 76426-788 76426-666 76426-820 76426-786 76426-664 76426-708 76426-828 76426-706 76426-826 76430-542 76430-544 76430-546 76430-548 76431-308 76426-762 76426-640 76426-760 76430-540 76426-638 76426-758 76426-636 76426-756 76426-634 76426-754 76426-632 76426-752 76426-598 76426-652 76426-772 76426-650 76426-770 76426-802 76426-648 76426-768 76426-800 76426-646 76426-766 76426-644 76426-764 76426-642 76426-808 76426-806 76426-804
Avantor® Hichrom HI-FFAP EXT GC Columns
Chromatography Columns
Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols, with extended temperature range.
  • Phase composition: Extended temperature crossbonded nitroterephthalic-acid-modified polyethylene glycol (PEG)
  • Long lifetime
Max temperature limits may change depending on the film thickness.