Avantor® Hichrom HI-624 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI08-53-100-60 HI08-32-010-60 HI08-18-020-50 HI08-18-018-60 HI08-10-020-15 HI082502560G5 HI08-53-050-15 HI08-32-100-10 HI08-53-100-75 HI08-25-010-50 HI08-25-300-30 HI08-05-005-5 HI08-10-020-10 HI08-25-100-50 HI08-32-010-30 HI08-53-300-75 HI0832010105 HI08-10-010-10 HI08-25-050-15 HI08-53-050-60 HI0825050105 HI08-18-100-20 HI08-53-010-30 HI08-18-040-50 HI08-32-025-15 HI08-18-040-20 HI08-53-300-30 HI08-10-040-15 HI08-25-100-10 HI08-18-020-40 HI08-32-010-15 HI08-32-100-30 HI08-53-025-15 HI08-32-100-15 HI08-53-300-60 HI08-25-140-30 HI08-32-150-60 HI08-15-084-30 HI08-18-040-10 HI08-32-050-50 HI08-25-025-30 HI08-53-300-15 HI08-32-300-30 HI08-53-025-50 HI08-53-050-10 HI08-15-015-15 HI08-53-300-50 HI08-20-112-50 HI083218030G5 HI08-53-150-15 HI08-32-050-60 HI0825140105 HI08-18-020-20 HI082502530G5 HI08-32-050-30 HI08-32-300-15 HI08-25-025-15 HI08-53-150-60 HI08-25-010-60 HI08-32-180-60 HI08-32-025-50 HI08-15-015-30 HI08-18-100-50 HI08-15-084-20 HI08-25-010-15 HI08-10-010-5 HI08-20-112-60 HI0832180105 HI08-15-084-15 HI085330030G5 HI08-25-050-60 HI08-25-025-10 HI08-53-100-30 HI08-53-300-10 HI08-53-150-50 HI08-32-050-15 HI08-32-150-30 HI08-15-015-10 HI08-53-025-60 HI08-32-300-10 HI08-18-100-40 HI085310030G5 HI08-20-112-20 HI08-10-010-15 HI08-25-100-60 HI08-25-140-50 HI08-32-050-10 HI08-32-025-30 HI08-10-020-5 HI08-32-150-10 HI08-32-100-60 HI08-32-180-50 HI08-18-018-20 HI08-25-050-10 HI08-25-100-30 HI082514060G5 HI08-18-018-50 HI08-18-018-10 HI08-53-025-30 HI08-32-025-60 HI08-32-180-30 HI08-20-112-40 HI08-53-100-50 HI08-53-100-10 HI08-25-300-50 HI08-15-084-40 HI0832050105 HI08-53-150-10 HI08-32-300-60 HI08-32-150-15 HI08-32-010-50 HI0832100105 HI08-53-010-10 HI08-25-050-50 HI082502515G5 HI08-25-300-60 HI08-25-050-30 HI08-18-018-40 HI08050052.5 HI08-25-140-60 HI08-32-025-10 HI08-05-005-2 HI08-32-010-10 HI08-18-100-60 HI08-10-040-10 HI08-32-180-75 HI08-25-100-15 HI08-53-150-75 HI08-15-015-5 HI08-15-015-20 HI08-25-010-30 HI08-10-040-5 HI082514030G5 HI0832300105 HI08-53-050-30 HI08-18-040-40 HI08-32-100-50 HI08-53-100-15 HI08-32-150-50 HI08-53-150-30 HI08-18-020-60 HI08-53-010-15 HI08-25-025-60 HI0832150105 HI08-18-020-10 HI08-32-300-50 HI08-25-025-50 HI0825025105 HI08-53-050-50 HI0832025105
76426-184EA 1659.42 USD
76426-184 76426-080 76425-986 76425-978 76430-468 76431-264 76426-168 76426-106 76426-186 76426-018 76426-064 76430-458 76430-466 76426-052 76426-076 76426-210 76426-072 76430-460 76426-038 76426-174 76426-036 76425-998 76426-156 76425-996 76426-086 76425-992 76426-204 76430-474 76426-046 76425-984 76426-074 76426-112 76426-158 76426-110 76426-208 76426-058 76426-128 76430-492 76425-990 76426-102 76426-028 76426-202 76426-146 76426-162 76426-166 76430-480 76426-206 76426-010 76431-270 76426-190 76426-104 76426-056 76425-982 76431-262 76426-100 76426-144 76426-026 76426-196 76426-020 76426-136 76426-090 76430-484 76426-002 76430-490 76426-014 76430-464 76426-012 76426-130 76430-488 76431-274 76426-044 76426-022 76426-180 76426-200 76426-194 76426-098 76426-124 76430-478 76426-164 76426-140 76426-000 76431-272 76426-006 76430-462 76426-054 76426-060 76426-094 76426-088 76430-470 76426-118 76426-116 76426-134 76425-972 76426-034 76426-050 76431-268 76425-976 76425-970 76426-160 76426-092 76426-132 76426-008 76426-182 76426-176 76426-066 76430-494 76426-096 76426-188 76426-150 76426-122 76426-078 76426-108 76426-152 76426-042 76431-260 76426-068 76426-040 76425-974 76430-456 76426-062 76426-082 76430-454 76426-070 76426-004 76430-472 76426-138 76426-048 76426-198 76430-486 76430-482 76426-016 76430-476 76431-266 76426-142 76426-170 76425-994 76426-114 76426-178 76426-126 76426-192 76425-988 76426-154 76426-032 76426-120 76425-980 76426-148 76426-030 76426-024 76426-172 76426-084
Avantor® Hichrom HI-624 GC Columns
Chromatography Columns
General purpose low-to-mid-polarity phase for a wide range of applications, including (for example): Residual solvents, volatile pollutants.
  • Phase composition: Crossbonded 6% cyanopropylphenyl, 94% Methyl polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.