Avantor® Hichrom HI-50 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI3532020105 HI35-20-020-50 HI3532020100 HI35-32-020-60 HI35-25-025-50 HI35-32-020-50 HI35-32-025-60 HI35-32-025-50 HI3520010105 HI3525025100 HI35-20-010-20 HI3525025105 HI3520020100 HI3525020100 HI35-20-010-50 HI3532025105 HI3520020105 HI35-20-020-60 HI35-25-025-60 HI35-32-025-30 HI35-20-010-10 HI3520010100 HI35-20-020-20 HI35-25-020-30 HI35-32-020-30 HI35-20-010-60 HI35-20-010-40 HI35-18-010-10 HI3525020105 HI35-25-025-30 HI3532025100 HI35-20-020-40 HI35-18-010-20 HI35-25-020-60 HI35-25-020-50 HI35-18-010-40
76425-916EA 932.5 USD
76425-916 76430-444 76425-918 76430-446 76430-448 76425-874 76425-896 76425-910 76425-898 76425-876 76425-912 76425-878 76425-914 76425-880 76425-882 76425-860 76425-862 76425-884 76425-904 76425-906 76425-908 76425-864 76425-886 76425-866 76425-888 76425-920 76430-450 76425-868 76425-900 76430-452 76425-902 76425-890 76425-892 76425-870 76425-894 76425-872
Avantor® Hichrom HI-50 GC Columns
Chromatography Columns
High polarity phase, ideal for detailed FAMEs and Dioxins isomers separation.
  • Phase composition: 50% Cyanopropyl, 50% Methyl Polysiloxane
Max temperature limits may change depending on the film thickness.