Avantor® Hichrom HI-35 GC columns
Supplier: Avantor®
Avantor® Hichrom
HI34-32-025-15 HI34-25-050-30 HI34-53-010-10 HI34-10-040-10 HI34-32-150-30 HI3432050105 HI34-25-025-60 HI34-53-025-15 HI34-25-025-30 HI34-32-025-10 HI34-32-050-60 HI34-32-150-60 HI3432100105 HI34-53-100-15 HI34-15-015-5 HI34-53-300-10 HI34-53-010-15 HI34-25-025-50 HI34-15-015-30 HI34-53-300-75 HI34-32-100-30 HI34-53-025-50 HI34-18-040-60 HI34-32-025-60 HI3432025105 HI34-53-150-75 HI34-53-025-30 HI34-20-020-40 HI34-32-050-15 HI34-25-050-15 HI34-10-040-5 HI34-20-020-60 HI34-10-010-15 HI34-20-100-10 HI34-10-020-15 HI3425050105 HI34-25-025-10 HI34-32-010-50 HI34-32-025-30 HI34-20-040-40 HI34-53-265-30 HI34-20-010-60 HI34-20-010-20 HI34-53-100-60 HI34-53-300-15 HI34-53-050-15 HI34-25-100-30 HI3425025105 HI34-32-010-30 HI34-18-018-40 HI34-05-005-5 HI34-53-150-10 HI34-53-100-30 HI34-25-010-15 HI34-20-020-50 HI34-18-040-20 HI34-10-020-10 HI34-32-150-10 HI34-15-015-10 HI34-25-100-10 HI34-10-040-15 HI34-53-100-10 HI34-18-020-10 HI34-53-050-10 HI34-32-025-50 HI34-18-018-50 HI3425100105 HI34-53-050-50 HI34-32-300-60 HI34-25-050-60 HI34050052.5 HI34-32-100-15 HI34-10-010-10 HI34-20-020-20 HI345310030G5 HI34-25-050-50 HI34-32-050-50 HI34-18-020-20 HI34-53-300-60 HI34-25-100-50 HI34-32-010-15 HI34-53-300-30 HI34-20-100-20 HI34-53-050-60 HI34-32-050-10 HI34-53-150-30 HI34-32-100-10 HI34-10-020-5 HI34-32-010-60 HI34-53-150-50 HI34-10-010-5 HI34-20-010-40 HI34-18-018-60 HI34-05-005-2 HI34-53-010-30 HI34-25-010-30 HI34-32-300-30 HI34-20-040-20 HI3432010105 HI34-20-010-50 HI34-53-050-30 HI34-32-300-50 HI34-18-040-50 HI34-18-018-20 HI34-15-015-15 HI34-53-100-75 HI34-18-020-50 HI34-53-150-15 HI34-32-100-60 HI34-15-015-20 HI3432300105 HI34-18-040-10 HI34-20-100-40 HI34-32-150-50 HI34-32-300-15 HI34-53-265-10 HI34-25-010-50 HI34-32-010-10 HI34-25-100-15 HI34-25-010-60 HI34-20-100-60 HI34-25-025-15 HI34-20-040-50 HI34-32-150-15 HI34-20-020-10 HI34-20-010-10 HI34-18-020-60 HI34-53-100-50 HI342502530G5 HI34-32-300-10 HI34-25-050-10 HI3432150105 HI34-53-300-50 HI34-18-020-40 HI34-18-018-10 HI342502560G5 HI34-20-040-10 HI34-32-050-30 HI34-53-025-60 HI34-18-040-40 HI34-53-150-60 HI34-32-100-50 HI342502515G5 HI34-25-100-60 HI34-20-040-60
76425-816EA 1497.85 USD
76425-816 76425-818 76425-776 76425-654 76425-778 76425-656 76425-810 76425-812 76425-658 76425-814 76425-660 76425-780 76425-662 76425-782 76425-664 76425-784 76425-828 76425-706 76425-708 76425-666 76425-820 76430-392 76425-786 76425-822 76425-668 76430-394 76425-788 76425-700 76425-824 76425-702 76430-396 76425-826 76425-704 76430-398 76425-790 76431-144 76425-792 76431-142 76425-670 76425-794 76425-672 76431-140 76425-796 76430-390 76425-674 76431-146 76425-754 76425-632 76425-756 76425-634 76425-758 76425-636 76425-638 76425-640 76425-760 76425-642 76425-762 76425-806 76425-808 76425-644 76425-764 76425-646 76425-800 76425-766 76425-802 76425-648 76425-768 76425-804 76425-770 76425-772 76425-650 76425-774 76425-652 76430-400 76425-618 76430-402 76430-404 76430-406 76425-732 76425-698 76425-852 76425-734 76425-854 76425-612 76425-736 76425-856 76425-614 76425-738 76425-858 76425-616 76425-620 76425-740 76425-622 76425-742 76425-624 76425-744 76425-626 76425-746 76425-628 76425-748 76425-750 76425-752 76425-630 76425-718 76430-388 76425-838 76425-798 76430-380 76425-710 76425-830 76425-676 76425-712 76430-382 76425-832 76425-678 76430-384 76425-714 76425-834 76430-386 76425-716 76425-836 76425-680 76425-682 76425-684 76425-686 76425-840 76430-378 76425-728 76425-842 76425-688 76425-720 76425-844 76425-722 76425-846 76425-724 76425-848 76430-376 76425-726 76425-690 76425-692 76425-694 76425-730 76425-696 76425-850 76430-408
Avantor® Hichrom HI-35 GC columns
Chromatography Columns
Really versatile general purpose mid-polarity phase for a wide range of applications. Ideal in confirmation analysis.
  • Phase composition: Crossbonded Silphenylene - 35% Phenyl - 65% Methyl Polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.