Avantor® Hichrom HI-225 GC Columns
Supplier: Avantor®
Avantor® Hichrom
HI33-32-025-10 HI33-20-010-10 HI33-20-010-50 HI33-10-010-15 HI33-32-010-10 HI33-18-018-20 HI33-15-015-15 HI33050052.5 HI33-32-050-50 HI33-25-050-50 HI33-20-040-20 HI33-53-010-15 HI33-32-025-50 HI33-18-040-20 HI33-32-050-60 HI33-32-025-60 HI33-18-020-20 HI33-20-020-50 HI3325050105 HI33-20-040-10 HI33-53-050-30 HI33-25-010-30 HI33-18-018-10 HI33-25-010-15 HI33-10-020-15 HI33-18-040-40 HI33-15-015-30 HI33-32-025-15 HI33-20-100-20 HI33-10-020-10 HI33-15-015-5 HI33-32-025-30 HI33-32-100-10 HI33-32-100-50 HI33-25-025-50 HI33-53-050-50 HI33-20-010-20 HI33-10-010-10 HI33-05-005-2 HI33-25-010-60 HI33-53-010-30 HI33-25-010-50 HI3332100105 HI33-25-025-10 HI33-25-050-15 HI33-32-100-30 HI33-15-015-20 HI3332025105 HI33-25-050-60 HI33-32-010-60 HI33-32-050-10 HI33-32-010-15 HI33-32-100-15 HI33-20-100-10 HI33-18-018-50 HI33-53-010-10 HI33-20-100-40 HI33-25-050-30 HI33-10-020-5 HI33-20-040-40 HI33-53-025-60 HI33-20-020-10 HI33-32-010-50 HI33-32-010-30 HI33-53-025-15 HI33-25-025-60 HI33-53-050-60 HI33-10-040-5 HI33-20-020-20 HI33-18-040-10 HI33-15-015-10 HI33-53-050-15 HI33-25-025-15 HI33-20-020-60 HI33-32-100-60 HI33-32-050-15 HI3332050105 HI33-20-100-60 HI33-20-100-50 HI33-53-050-10 HI33-10-010-5 HI33-18-018-40 HI3325025105 HI33-25-050-10 HI33-25-025-30 HI33-20-020-40 HI33-18-020-40 HI33-20-040-50 HI33-20-010-60 HI33-32-050-30 HI33-53-025-50 HI33-20-040-60 HI33-18-040-50 HI33-18-020-10 HI33-05-005-5 HI33-10-040-10 HI33-18-040-60 HI33-18-018-60 HI33-20-010-40 HI3332010105 HI33-53-025-30 HI33-10-040-15
76425-534EA 870.84 USD
76425-534 76425-536 76425-538 76425-540 76425-542 76425-544 76425-546 76425-548 76425-550 76425-552 76425-554 76430-344 76430-346 76430-348 76425-512 76425-478 76425-514 76425-516 76430-342 76425-518 76425-482 76425-484 76425-486 76425-488 76425-520 76425-480 76425-522 76425-524 76425-526 76425-528 76425-492 76425-494 76425-530 76425-496 76425-532 76425-498 76425-490 76430-366 76430-368 76425-578 76425-610 76425-456 76430-360 76425-458 76430-362 76430-364 76425-460 76425-580 76425-462 76425-582 76425-464 76425-584 76425-466 76425-586 76425-508 76430-356 76430-358 76425-468 76425-500 76425-588 76430-350 76425-502 76425-504 76430-352 76430-354 76425-506 76425-592 76425-470 76425-594 76425-472 76425-596 76425-474 76425-510 76425-598 76425-476 76425-590 76425-556 76425-558 76425-560 76425-442 76425-562 76425-444 76425-564 76425-608 76425-600 76425-446 76430-370 76425-566 76425-448 76425-602 76430-372 76425-568 76425-604 76430-374 76425-606 76425-570 76425-572 76425-450 76425-574 76425-452 76425-576 76425-454
Avantor® Hichrom HI-225 GC Columns
Chromatography Columns
General purpose mid-to-high-polarity phase for a wide range of applications, like, for example: FAMEs, carbohydrates, sterols, flavor compounds.
  • Phase composition: Crossbonded 25% Cyanopropyl, 25% Phenyl, 50% Methyl Polysiloxane
Max temperature limits may change depending on the film thickness.