Avantor® Hichrom HI-200 GC columns
Supplier: Avantor®
Avantor® Hichrom
HI32-53-050-60 HI32-32-010-15 HI32-32-150-30 HI32-20-010-50 HI32-10-020-10 HI32-20-010-20 HI32-53-150-10 HI32-32-050-30 HI32-53-100-60 HI32-32-025-15 HI32-18-040-60 HI32-25-010-15 HI32-10-040-5 HI32-10-020-5 HI32-10-010-10 HI32-32-100-10 HI32-32-010-10 HI32-15-015-10 HI32-25-010-60 HI32-32-025-60 HI32-25-025-50 HI32-53-050-15 HI32-32-010-30 HI32-53-100-75 HI32-32-025-50 HI32-25-025-60 HI32-53-100-30 HI32-20-100-20 HI32-32-050-60 HI32-25-025-15 HI3225050105 HI32-15-015-20 HI32-18-020-10 HI32-32-100-30 HI32-25-100-10 HI32-20-020-20 HI32-18-020-40 HI32-53-025-60 HI32-53-150-75 HI32-10-010-15 HI32050052.5 HI32-18-018-40 HI32-32-150-10 HI32-25-100-50 HI3225025105 HI32-32-050-15 HI32-10-040-15 HI32-53-150-50 HI32-32-050-10 HI32-05-005-5 HI32-20-040-60 HI32-53-100-15 HI32-20-100-10 HI32-20-040-20 HI32-10-020-15 HI32-20-020-50 HI32-10-010-5 HI32-53-150-60 HI32-10-040-10 HI32-53-050-30 HI32-18-040-10 HI32-18-018-10 HI3232100105 HI32-25-100-60 HI32-20-040-40 HI32-20-010-10 HI32-53-150-15 HI32-25-050-15 HI32-15-015-5 HI32-32-100-15 HI32-25-025-10 HI32-25-010-30 HI32-32-025-30 HI32-53-010-30 HI32-15-015-30 HI32-20-020-40 HI32-32-150-60 HI32-18-018-20 HI32-32-150-50 HI32-25-050-60 HI32-32-010-60 HI32-53-100-10 HI3232010105 HI32-53-025-15 HI32-25-050-30 HI32-25-100-30 HI32-20-010-60 HI32-05-005-2 HI32-25-100-15 HI32-53-050-50 HI32-32-150-15 HI32-20-040-50 HI32-53-010-15 HI32-18-018-50 HI32-53-010-10 HI32-53-025-30 HI32-53-050-10 HI32-20-020-10 HI32-32-100-50 HI32-32-050-50 HI32-53-025-50 HI3232050105 HI32-18-040-40 HI32-20-100-60 HI32-25-050-50 HI32-20-010-40 HI32-18-020-20 HI32-18-018-60 HI32-25-010-50 HI32-25-025-30 HI32-20-040-10 HI32-18-040-50 HI32-18-040-20 HI32-53-100-50 HI32-53-150-30 HI3232150105 HI32-32-010-50 HI32-25-050-10 HI32-20-100-50 HI3232025105 HI3225100105 HI32-20-020-60 HI32-15-015-15 HI32-32-025-10 HI32-20-100-40 HI32-32-100-60
76430-322EA 925.14 USD
76430-322 76430-324 76430-326 76430-328 76425-412 76425-258 76425-414 76425-416 76430-320 76425-418 76425-262 76425-382 76425-264 76425-384 76425-420 76425-266 76425-386 76425-422 76425-268 76425-388 76425-300 76425-260 76425-380 76430-312 76430-314 76430-316 76430-318 76425-424 76425-302 76425-426 76425-304 76425-428 76425-306 76425-308 76430-310 76425-394 76425-272 76425-396 76425-274 76425-398 76425-310 76425-430 76425-276 76425-312 76425-278 76425-432 76430-308 76425-390 76425-392 76425-270 76425-358 76425-236 76425-238 76430-340 76425-240 76425-360 76425-242 76425-362 76425-244 76425-364 76425-400 76425-246 76425-366 76430-334 76430-336 76430-338 76425-248 76425-402 76425-368 76425-404 76425-406 76430-330 76425-408 76430-332 76425-372 76425-250 76425-374 76425-252 76425-376 76425-254 76425-378 76425-256 76425-410 76425-370 76425-336 76425-338 76425-340 76425-342 76425-224 76425-344 76425-226 76425-346 76425-228 76425-348 76425-350 76425-352 76425-230 76425-354 76425-232 76425-356 76425-234 76425-314 76425-434 76425-316 76425-436 76425-318 76425-438 76425-284 76425-440 76425-286 76425-288 76425-320 76425-322 76425-280 76425-282 76425-324 76425-326 76425-328 76425-294 76425-330 76425-296 76425-332 76425-298 76425-334 76425-290 76425-292
Avantor® Hichrom HI-200 GC columns
Chromatography Columns
Unique selectivity trifluoropropyl methyl polysiloxane stationary phase.
  • Phase composition: Crossbonded trifluoropropyl methyl polysiloxane
  • Long lifetime
Max temperature limits may change depending on the film thickness.