White Diamond Series Stainless Steel Freezer, Norlake Scientific
Supplier: Horizon Scientific
NSWDF231SSS0A3
76422-120EA 7883 USD
76422-120
White Diamond Series Stainless Steel Freezer, Norlake Scientific
Freezers Laboratory / General Purpose Freezers
White diamond series glass door laboratory and medical stainless steel freezer with microprocessor temperature controller. This unit features hydrocarbon natural refrigerants and a touch screen digital temperature display with user friendly interface. Audible and visual high/low alarms and remote alarm contacts are some of the features of this well rounded unit. Operating temperature to  (–30 °C).
  • Natural refrigerant
  • Keyed door lock
  • Probe access port
  • Integrated door handle
Certificaciones: ETL Listed
Precaución: Auto defrost freezers incorporate an electric heater installed on the evaporator that assists in the removal of the frost and ice. Although the heater helps speed up the frost and ice removal, it can raise the temperatures within the freezer.