VWR® E-Series Balances with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-3002E/C VWR-3001E/C VWR-10001E/C VWR-20001E/C VWR-6001E/C VWR-1002E/C
76378-452EA 1546.91 USD
76378-452 76378-454 76378-458 76378-350 76378-456 76378-436
VWR® E-Series Balances with Calibration Certificate
Balances and Scales
The VWR® E-Series/C balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Backlit, LCD screen
  • External calibration
  • RS232 communication interface
  • Five year warranty
The portable balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These economical and versatile balances offer a variety of standard features, such as multiple weighing units, sturdy construction, simple operation, and quick stabilization time.
The E-series has external calibration and requires purchase of mass standards to perform calibration procedure.
Certificaciones: ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate; CE marked.
Información para pedidos: The VWR-10001E and VWR-20001E models feature an internal rechargeable battery. All other units utilize 6×AA batteries. All units supplied with switching AC adapter, weighing pan, and user manual. Includes five-year warranty. Accessories not included.