VWR® B2T-Series Balances with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-6002B2T/C VWR-363B2T/C VWR-2002B2T/C VWR-310B2T/C VWR-603B2T/C VWR-60B2T/C VWR-6001B2T/C VWR-220B2T/C VWR-20001B2T/C VWR-160B2T/C VWR-10001B2T/C VWR-4502B2T/C VWR-1003B2T/C
76378-424EA 1543.96 USD
76378-424 76458-550 76378-422 76378-434 76378-420 76458-554 76378-432 76378-430 76458-552 76378-428 76378-418 76458-548 76378-426
VWR® B2T-Series Balances with Calibration Certificate
Balances and Scales
The VWR® B2T-series/C balances which include an ISO/IEC 17025:2017 calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Large 5" color touch screen
  • Touchless operation via IR proximity sensors
  • 2×RS232, Ethernet and USB (A and B) communication interfaces
  • Includes five-year warranty
  • Tool-free assembly and disassembly of the three-door draft shield
The balances include a certificate of calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights.
These user-friendly balances provide high quality results and offer a variety of standard features, such as a color touchscreen, GLP procedures, 17 weighing units, adjustable ambient condition settings, 11 application modes, and quick stabilization time.
The B2T-series balances have external calibration and requires purchase of mass standards to perform calibration procedure.
The 0.1 mg readability models feature a fixed 3-door draft shied (which can't be removed). The 0.001 g readability models feature a removable, glass draft shield.
Certificaciones: CE marked.
Información para pedidos: All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Corresponding models with internal calibration are also available, please see vwr.com for details. Accessories not included.