VWR® P2-Series Balances with Calibration Certificate
Supplier: VWR, part of Avantor
VWR®
VWR-20001P2H/C VWR-2002P2.H/C VWR-6001P2.H/C VWR-2002P2HC/C VWR-2002P2/C VWR-30000P2H/C VWR-203P2/C VWR-10001P2H/C VWR-60000P2H/C VWR-3101P2/C VWR-602P2/C VWR-6001P2HC/C
76480-264EA 1612.06 USD
76480-264 76378-372 76378-362 76378-370 76480-266 76480-268 76480-270 76378-368 76378-366 76378-374 76480-262 76378-364
VWR® P2-Series Balances with Calibration Certificate
Balances and Scales
VWR® P2-Series/C balances which include an ISO/IEC 17025:2017 Calibration Certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Internal, rechargeable battery
  • Backlit, LCD screen
  • External and internal calibration options
  • RS232 and USB (A and B) communication interfaces
  • Five year warranty
The economical balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These compact, portable balances offer a variety of standard features, such as a multiple weighing modes, advanced communication features, multiple weighing units, and quick stabilization time.
The P2 series features USB (A and B) and RS232 interfaces. The VWR-30000P2.H and VWR-60000P2.H models only feature a RS232 connection.
Certificaciones: ISO/IEC 17025:2017 Accredited Calibration and Testing Laboratory issues Calibration Certificate; CE marked
Información para pedidos: All units supplied with switching AC adapter, weighing pan, user manual, and quick start guide. Includes five-year warranty. Accessories not included.