VWR® Pressure Sensitive Color-Coded Labeling Tapes
Supplier: VWR, part of Avantor
VWR®
VWR-T25-15 VWR-T24-25 VWR-T24-22 VWR-T15-13 VWR-T25-20 VWR-T14-24 VWR-T24-24 VWR-T25-22 VWR-T25-58 VWR-T35-19 VWR-T35-22 VWR-T25-17 VWR-T14-21 VWR-T15-21 VWR-T15-15 VWR-T35-9 VWR-T24-20 VWR-T35-25 VWR-T15-12 VWR-T15-26 VWR-T25-25 VWR-T25-14 VWR-T14-58 VWR-T15-8 VWR-T15-10 VWR-T34-18 VWR-T25-19 VWR-T25-9 VWR-T34-20 VWR-T34-21 VWR-T25-21 VWR-T15-17 VWR-T25-26 VWR-T35-21 VWR-T15-16 VWR-T24-21 VWR-T35-15 VWR-T15-22 VWR-T14-20 VWR-T15-20 VWR-T35-58 VWR-T14-19 VWR-T35-24 VWR-T34-25 VWR-T15-18 VWR-T15-58 VWR-T34-22 VWR-T15-9 VWR-T35-8 VWR-T34-24 VWR-T25-24 VWR-T15-25 VWR-T25-18 VWR-T24-18 VWR-T35-18 VWR-T35-26 VWR-T35-17 VWR-T15-14 VWR-T34-58 VWR-T35-20 VWR-T34-19 VWR-T35-12 VWR-T15-24 VWR-T14-22 VWR-T35-10 VWR-T15-11 VWR-T24-58 VWR-T24-19 VWR-T14-25 VWR-T14-18 VWR-T15-19
76376-520CS 23.44 USD
76376-520 76376-514 76376-510 76376-466 76376-528 76376-454 76376-512 76376-532 76376-540 76376-570 76376-576 76376-522 76376-450 76376-484 76376-472 76376-588 76376-506 76376-580 76376-464 76376-492 76376-536 76376-518 76376-458 76376-498 76376-460 76376-544 76376-526 76376-542 76376-548 76376-550 76376-530 76376-476 76376-538 76376-574 76376-474 76376-508 76376-564 76376-486 76376-448 76376-482 76376-584 76376-446 76376-578 76376-556 76376-478 76376-494 76376-552 76376-500 76376-586 76376-554 76376-534 76376-490 76376-524 76376-502 76376-568 76376-582 76376-566 76376-468 76376-558 76376-572 76376-546 76376-562 76376-488 76376-452 76376-560 76376-462 76376-516 76376-504 76376-456 76376-444 76376-480
VWR® Pressure Sensitive Color-Coded Labeling Tapes
Tapes
Can be written on using a pen, pencil, and/or a permanent marker without smearing. Can be adhered, removed, and reapplied without leaving behind the adhesive residue.
  • Oil-resistant
  • Acid-resistant
  • Water-resistant
  • Withstands temperatures ranging from −23 to 121 °C
Common in laboratories for color-coding or labeling.