PD-Tip™ II Precision Dispenser Tips, BrandTech® Scientific, Inc.
Supplier: BrandTech
PD-Tips II
705716 705744 705746 705732 705736 705714 705748 705734 705700 705738 705710 705730 705702 705740 705718 705742 705706 705708 705712 705704 705720
76338-304PK 57.68 USD
76338-304 76338-268 76338-302 76338-308 76338-306 76338-280 76338-292 76338-270 76338-290 76338-284 76338-296 76338-274 76338-282 76338-294 76338-272 76338-288 76338-278 76338-300 76338-286 76338-298 76338-276
PD-Tip™ II Precision Dispenser Tips, BrandTech® Scientific, Inc.
Pipet Tips
These dispenser tips fit all repeating pipettes using standard tips, such as BRAND® HandyStep®, Rainin® AutoRep™, Gilson® Repetman® and Eppendorf® 4780.
  • Precision-molded of premium polypropylene and high density polyethylene (PP/LCP for 0.1 ml size)
  • Tip size automatically recognized by BRAND® HandyStep® touch, HandyStep® touch S and HandyStep® electronic pipettes
  • BIO-CERT® sterile tips are free of DNA, RNase, ATP and endotoxin within detectable limits (batch certificate included) to meet the most stringent sample integrity requirements
Available in ten sizes (from 0.1 to 50 ml), both non sterile and BIO-CERT® sterile.
Compatible with most repeater pipettes including the BRAND HandyStep® touch, HandyStep® touch S, HandyStep® S and HandyStep® electronic, Rainin® AutoRep™ E, AutoRep™ S and AutoRep™ M, Gilson® Repetman®, and all other repeating pipettes using standard tips. Tip size is automatically recognized by BRAND® HandyStep® touch, HandyStep® touch S, HandyStep® electronic, Rainin® AutoRep™ E and Gilson® Repetman®.
High quality materials: manufactured entirely from high-quality polyethylene and polypropylene (piston of 0.1 ml tip is liquid crystal polymer-LCP).
*Recent model Eppendorf® Repeater™ instruments feature a proprietary design to preclude their use with tips from other manufacturers.
Certificaciones: BIO-CERT® PD-Tip™ II tips are supplied with batch certificate to document accuracy and precision testing and help simplify ISO/GLP compliance.
Información accesorios: 25 ml and 50 ml PD-Tip™ II dispenser tips require the use of an adapter. One is supplied per box; additional adapters are available non sterile, or BIO-CERT® sterile.
Envase: Non sterile PD-Tip™ II tips are packaged bulk in recyclable packaging. BIO-CERT® tips are individually-wrapped, also in recyclable packaging.