VWR® Benchtop Cryogenic Racks
Supplier: VWR, part of Avantor
VWR®
5610-40-03AV1 5610-43-03AV1
76337-370 76337-372
VWR® Benchtop Cryogenic Racks
Racks Cryogenic Vial Racks
These reversible benchtop chillers hold either 0.5 or 0.65 ml tubes on one side and 1.5 to 2.0 ml tubes on the other.
  • Constructed of polycarbonate
  • Can maintain cold temperatures for up to four hours
  • Non-toxic gel fill
Racks are available in two temperature ranges and can be frozen overnight for maximum duration.