BRAND® Inserts 2 in 1 for Cell Culture, BrandTech® Scientific, Inc.
Supplier: BrandTech
Brand®
782716 782741 782717 782731 782706 782757 782721 782710 782751 782700 782746 782737 782730 782720 782740 782727 782736 782756 782750 782707 782747 782701 782711 782726
76337-192PK 439.24 USD
76337-192 76337-194 76337-190 76337-188 76337-232 76337-200 76337-244 76337-234 76337-246 76337-202 76337-224 76337-196 76337-240 76337-230 76337-186 76337-242 76337-198 76337-208 76337-236 76337-226 76337-204 76337-238 76337-206 76337-228
BRAND® Inserts 2 in 1 for Cell Culture, BrandTech® Scientific, Inc.
Microplates Insert Plates and Inserts
BRAND® insert 2 in 1 offers unlimited flexibility for 2D and 3D cell culture. Inserts can be used as a free standing or hanging insert for co-culture, air-lift cultures, migration assays and more.
  • cellGrade™ plus surface for enhanced cell attachment
  • PC or PET membrane
  • 0.4 or 8 µm pore sizes
  • Compatible with all common 6-, 12-, or 24-well plates and tissue culture dishes
Common applications include co-culture, trans-migration, chemotaxis, transport/barrier analysis (TEER), cytotoxicity and air-lift culture assays.
Pyrogen and endotoxin free according to LAL test, detection limit 0.01 EU/ml.
Certificaciones: Manufactured in a Class 8 cleanroom, in accordance with ISO 14644-1. Sterile according to ISO 11137, SAL 10-6. Non-cytoxic according to ISO 10993-5.
Envase: Inserts are available individually packed or in multi-packs.
Precaución: These products can expose you to chemicals including α-Methyl styrene (alpha-Methylstyrene), which is known to cause cancer, and Di-n-butyl Phthalate (DBP), which is known to cause birth defects or other reproductive harm.