VWR® LTS Compatible Adjustable Volume Pipettors
Supplier: VWR, part of Avantor
76382-704 76382-700 76335-572 76382-696 76335-748 76335-568 76335-746 76382-698 76335-742 76382-702 76335-744 76335-570 76382-708 76382-694 76382-706
76382-698EA 1301.35 USD
76382-698 76335-572 76382-696 76382-694 76335-742 76382-706 76382-704 76382-702 76382-700 76335-570 76382-708 76335-568 76335-744 76335-746 76335-748
VWR® LTS Compatible Adjustable Volume Pipettors
Pipets Micropipets
Patented tip ejection technology reduces fatigue and makes the VWR LTS compatible pipettor a top choice to improve your lab’s ergonomics.
  • Calibrated to ISO 8655 standards with Biotix xTIP4 tips
  • Fully compatible with Rainin LTS tips
  • Light spring forces produce a smooth and controlled aspiration with perfect balance
  • Padded ejector button to reduce hand vibrations during tip ejection
  • Simple volume adjustment with a secure lock to prevent volume drift
  • Available in single, 8, and 12-channel models with a symmetrical design for ambidextrous use
The VWR LTS compatible pipettor uses durable and lightweight components that deliver a precise and accurate liquid transfer while maintaining optimal hand comfort.
Certificaciones: ISO 8655 certified. Calibration certificate included in the box with each pipette. Unique serial ID number under the finger hook for GLP/GMP compliance.
Precaución: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.