VWR® A-Series Balances Legal-for-Trade (NTEP)
Supplier: VWR, part of Avantor
VWR®
VWR-314AC.N VWR-224AC.N VWR-2102AC.N VWR-1003AC.N VWR-363AC.N
76329-408EA 4270.18 USD
76329-408 76329-406 76329-410 76329-404 76329-402
VWR® A-Series Balances Legal-for-Trade (NTEP)
Balances and Scales
The A-series balances with NTEP certification feature state-of-the-art technology to provide the highest level of accuracy.
  • Class I and II options
  • Advanced features for weighing and documentation
  • Internal calibration
  • 2× RS232, USB (A and B), and Wi-Fi communication interfaces
  • Easy-to-read, adjustable, backlit display
The VWR® A-Series Balances with NTEP Certification offer the highest level or precision in legal-for-trade applications. These Class I and Class II certified balances feature an NTEP Certificate of Conformance indicating the devices meet the requirements of Handbook 44. Featuring internal calibration, a multitude of communication options, and built in data protections, the VWR A-Series balances provide advanced features for any application.
The large, back-lit LCD display allows for easy viewing of the weighing results. Data safety is provided via the internal memory allowing for recording up to 100000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. The VWR® A-Series Balances with NTEP Certification are the answer for organizations demanding precision and performance in legal-for-trade applications.
Certificaciones: NTEP certified for legal-for-trade applications; CE marked
Información accesorios: PRNT-01 (10799-132): Printer
AVBP-01 (10799-162): Granite platform
Información para pedidos: The 0.01 and 0.1 mg readability models feature a 3-door draft shield (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty. Accessories not included.
Precaución: Fragile, handle with care.