VITsafe™ Safety Wash Bottles, Brandtech
Supplier: BrandTech
V1352819 V1352869 V1352909 V1452829 V1352959 V1352899
76307-448PK 209.12 USD
76307-448 76307-446 76307-444 76307-368 76307-366 76307-364
VITsafe™ Safety Wash Bottles, Brandtech
Bottles Wash Bottles
Wide Mouth VITsafe™ safety wash bottles are made from low-density polyethylene (or Polypropylene for the Acetone bottle). Wash tubes are made of polypropylene.
  • Durable safety imprint in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Name in German, English, French, and Spanish
  • Printed with chemical formula, CAS No., hazard pictogram and signal word
  • Printed with risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Metal-free, color coded VENT-Caps reduce static overpressure, virtually eliminating leaks or drips from the dispensing tube. Wide-mouth openings simplify filling, even without a funnel.