VWR® B2T-Series Balances
Supplier: VWR, part of Avantor
VWR®
VWR-6001B2T VWR-310B2T VWR-1003B2T VWR-363B2T VWR-4502B2T VWR-6002B2T VWR-60B2T VWR-20001B2T VWR-10001B2T VWR-160B2T VWR-220B2T VWR-2002B2T VWR-603B2T
75870-328EA 1624.83 USD
75870-328 76458-480 76300-090 76300-086 76300-092 76300-094 76458-474 75870-332 75870-330 76458-476 76458-478 75870-334 76300-088
VWR® B2T-Series Balances
Balances and Scales
VWR® B2T-Series Balances combine advanced weighing features with a user-friendly touch display.
  • Reliable results
  • Multiple weigh modes
  • Large 5" color capacitive touch-screen display
  • Easy menu access and personalization to allow for customization of the balances home screen
  • Touchless operation via IR proximity sensors
  • Tool-free assembly and disassembly of the three-door draft shield
  • Five-year warranty
VWR® B2T-Series balances incorporate state-of-the-art features for advanced weighing applications at higher capacities.
These economical balances offer a variety of standard features such as external calibration, GLP procedures, 17 weighing units, color touch screen, multiple data interfaces, 11 application modes, and multilingual operation. Through the use of the built in proximity sensors, users are provided the opportunity to have touch-free operation by motioning over the sensors (printing results, zeroing, or taring). The VWR® B2T-Series provides high measurement precision and reliable results at an economical price.
Certificaciones: C-UL (US) and CE.
Información para pedidos: Calibration weight not included, must be purchased seperately. The 0.1 mg readability models feature a draft shied (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty. Accessories not included.