VWR® T-Series Balances
Supplier: VWR, part of Avantor
VWR®
VWR-363TC VWR-520TC VWR-225TC VWR-1003TC VWR-10102TC VWR-105TC VWR-603TC VWR-4502TC VWR-205TC VWR-3003TC VWR-310TC VWR-2102TC VWR-8102TC VWR-160TC VWR-220TC VWR-6002TC
76300-070EA 2865.24 USD
76300-070 76446-804 76300-056 76300-074 76300-082 76446-794 76300-072 76300-078 76446-796 76446-806 76446-802 76300-076 76446-808 76446-798 76446-800 76300-080
VWR® T-Series Balances
Balances and Scales
The VWR® T-Series balances feature state-of-the-art technology to provide the highest accuracy measurements.
  • Large, 5" color touch screen
  • Touchless operation via IR proximity sensors
  • Internal and external calibration
  • Seven databases
  • 2× RS232, USB (A and B), ethernet and Wi-Fi communication interfaces
  • Tool-free assembly and disassembly of the three-door draft shield
  • 5-year warranty
These balances provide accuracy and precision for any laboratory environment. Featuring internal calibration, IR proximity sensors for hands free operation, a multitude of communication options, and built in data protections, and provides advanced features for any application. The large, color touch screen allows for easy viewing of results and navigation through the balances menus. Data safety is provided via the internal memory allows for recording up to 500000 weighings. The analytical models feature a spacious 3-door draft shield that allows for tool-free assembly and disassembly of the weighing chamber. With its 5-year warranty, this balance will provide reliable results for years to come.
Certificaciones: C-UL (US) and CE.
Información para pedidos: Accessories not included (must be ordered separately).
Información de suministro: The 0.01 and 0.1 mg readability models feature a draft shield (shield can't be removed). The 0.001 g readability feature a removable, glass shield. All units supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Includes five-year warranty.