OmegaZen™ Pipette Controller, Argos Technologies®
Supplier: Argos Technologies
25300-98 25300-96 25300-97
76218-688EA 724.34 USD
76218-688 76218-690 76218-692
OmegaZen™ Pipette Controller, Argos Technologies®
Pipets Pipet Controllers
Precisely and rapidly pipet liquids with a comfortable, ergonomic design.
  • Easily fine-tune pipetting speed with thumbwheel control and speedometer display
  • Quick and quiet pump fills a 25 ml pipette in 3 seconds
  • Accepts glass or plastic serological pipettes, from 1 to 100 ml
  • Durable, UV-resistant plastic housing is ideal for use in cleanrooms and fume hoods
  • Avoid battery depletion with auto shut-off after 5 minutes of inactivity
  • Removable nosepiece and silicone adapter are autoclavable at 121 °C
Información accesorios: Four accessory filter options are available to choose from. All have a polypropylene housing and hydrophobic PTFE membrane. Filter should be replaced every three months, when the filter is contaminated, or suction capacity is slowed.
Información para pedidos: Each Pipette Controller includes two 0.45 µm sterile filters, nosepiece, silicone adapter, wall-mount holder, rechargeable lithium-ion battery, charging desk stand, and AC adapter.