VWR® ESD Micro Denier Scrub Pant
Supplier: VWR, part of Avantor
VWR®
2040ESW1NVY2XL 2040ESW1NVYS 2040ESW1NVY4XL 2040ESW1NVY5XL 2040ESW1NVY6XL 2040ESW1NVY1XL 2040ESW1NVYL 2040ESW1NVY3XL 2040ESW1NVYM 2040ESW1NVYXS
76202-682CS 1652.78 USD
76202-682 76202-696 76202-684 76202-698 76202-686 76202-700 76202-688 76202-690 76202-692 76202-680
VWR® ESD Micro Denier Scrub Pant
Workwear
This style of Scrub Pant is widely used in pharmaceutical cleanroom operations, and meets/exceeds the recommendations outlined in the IEST-RP-CC-003.4, Garment Considerations for Cleanrooms and Other Controlled Environments.
  • Combination elastic back and drawstring front for secure comfort
  • Leg opening with elastic back and three-position snaps for closure
  • Double-needle felled outer seams for extra durability
  • Garments include color-coded loops for easy size identification
  • All edges are pre-serged or laser cut for lint limitation
These scrubs provide an excellent base-layer of protection for any cleanroom garment system. Made with Sandwash material, this lightweight micro-denier fabric is woven from 100% continuous-filament polyester to eliminate any source for lint.