VWR® Classic Syringe Tips, Sterile
Supplier: VWR, part of Avantor
VWR®
ES40010-0008V ES40010-0009V ES40010-0012V ES40010-0011V ES40010-0013V ES40010-0010V
76163-600PK 360.86 USD
76163-600 76163-602 76163-608 76163-606 76163-610 76163-604
VWR® Classic Syringe Tips, Sterile
Pipet Tips
Sterile Classic Syringe Tips are individually wrapped, and certified to be pyrogen-, ATP-, Rnase-, and Dnase-free.
  • Ideal for viscous, infectious, and volatile liquids
  • Compatible with a variety of pipettors
Ripette®, Ripette®pro, Ripette®genX, eLineTM-Dispenser, Minilab 100, Minilab 101, Minilab 201, Distriman®, StepMate, EasyStep, EasyStep electronic, RepeatOne®, Handrop.
These syringe tips are not compatible with: HandyStep®, Repetman®.
They provide reliable and repeatable results for a wide range of liquids, including viscous, infectious, and volatile substances.