StarPlex Scientific Anaerobic Media in Glass Tube, with 2 Scored Rayon Swabs, Hardy Diagnostics
Supplier: Starplex Scientific
S120D
76045-448EA 637.38 USD
76045-448
StarPlex Scientific Anaerobic Media in Glass Tube, with 2 Scored Rayon Swabs, Hardy Diagnostics
Swabs Swabs in Tubes
The Anaerobic Transport System is a self-contained, sterile, ready to use system for the collection of specimens in sterile environments.
It includes a glass tube containing a semi-solid, pre-reduced transport medium, which is intended for maintaining the viability of a wide variety of microorganisms during transportation for up to 48 hours.
Fluids can be injected with a syringe directly into the tube through the pierceable, non-latex septum, limiting exposure to oxygen. The medium contains an indicator that will turn lavender if the anaerobic condition is compromised.