VWR® Polypropylene Measuring Cylinders, Class B
Supplier: VWR, part of Avantor
VWR®
345020 345040 345080 345050 345010 345070 345060 345030
76019-316CS 36.11 USD
76019-316 76019-318 76019-292 76019-258 76019-314 76019-254 76019-256 76019-294
VWR® Polypropylene Measuring Cylinders, Class B
Cylinders Measuring Cylinders
These VWR® Measuring Cylinders are manufactured from translucent polypropylene.
  • The cylinders feature a stable hexagonal base and a pouring spout
  • All cylinders also feature molded graduations for improved accuracy and consistency
When used continuously, the cylinders have a working temperature range of 0 to 121 °C (32 to 248 °F).
Certificaciones: Products are Class "B" as per DIN 12681/ISO 6706. Resin meets US FDA 21 CFR requirements.