EZwaste™ Replacement Fittings, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0942-OEM 330-0944-OEM 330-0940-OEM 330-0938-OEM 330-0932-OEM 330-0935-OEM 330-0939-OEM 330-0930-OEM 330-0950-OEM 330-0933-OEM 330-0936-OEM 330-0949-OEM 330-0931-OEM 330-0946-OEM 330-0934-OEM 330-0945-OEM 330-0948-OEM 330-0937-OEM 330-0947-OEM 330-0941-OEM
76018-662PK 22.45 USD
76018-662 76018-666 76018-536 76018-532 76018-520 76018-526 76018-534 76018-516 76018-678 76018-522 76018-528 76018-676 76018-518 76018-670 76018-524 76018-668 76018-674 76018-530 76018-672 76018-660
EZwaste™ Replacement Fittings, Foxx Life Sciences
Fittings
Replacement fittings and plugs provide the closed system necessary to prevent spills during HPLC liquid waste disposal. They are available in a variety of sizes and pack quantities.
  • Affordable solution for HPLC liquid waste disposal
  • Replacement fittings available in a variety of sizes
  • Variety of pack quantities available
  • Plugs available for unused ports
Foxx Life Sciences' replacement fittings and plugs secure waste tubes to carboys and cover any unused ports, providing the closed system necessary to prevent spills.
The EZwaste™ Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste™ Solvent Waste system also features a closed-loop design which protects against leaks and spills, improving overall lab safety.