EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste with Drum Adapter, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0C11-OEM 330-0C22-OEM 330-0C15-OEM 330-0C04-OEM 330-0C03-OEM 330-0C21-OEM 330-0C05-OEM 330-0C20-OEM 330-0C18-OEM 330-0C16-OEM 330-0C12-OEM 330-0C19-OEM 330-0C23-OEM
76018-628EA 384.02 USD
76018-628 76018-636 76018-626 76018-634 76018-624 76018-632 75805-422 75805-432 75805-430 76018-630 75805-428 75805-426 75805-424
EZwaste® UN/DOT 70 mm Cap Assembly for HPLC Solvent Waste with Drum Adapter, Foxx Life Sciences
Closures Bottle Closures
This cap system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb options for 1/4" or 3/8" ID
  • Drum adapter included
The EZwaste® UN/DOT Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste
Depending on application, carbon exhaust filter life cycle may vary.
Each system is manufactured to safely collect waste from up two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design which protects against leaks and spills, improving overall lab safety. The S-70 Caps with Drum Adapter come with a variety of port configurations for 1/16", 1/8", 1/4" O.D. tubing and hose barbs. All UN/DOT containers are rated for transport for easy waste pickup.