EZwaste® UN/DOT 51 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0A01-OEM 330-0A03-OEM
76018-606EA 331.36 USD
76018-606 76018-604
EZwaste® UN/DOT 51 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Closures Bottle Closures
This cap system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Tubing ports for 1/16" or 1/8" OD
The EZwaste® UN/DOT Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste
Depending on application, carbon exhaust filter life cycle may vary.
Each system is manufactured to safely collect waste from up to two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design which protects against leaks and spills, improving overall lab safety. The S-51 caps come with ports for either 1/16" O.D. tubing or 1/8" O.D. tubing. All UN/DOT containers are rated for transport for easy waste pickup.