EZwaste® XL 75 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Supplier: Foxx Life Sciences
332-6726-OEM 332-6713-OEM 332-6721-OEM 332-6725-OEM 332-6718-OEM 332-6730-OEM 332-6731-OEM 332-6720-OEM 332-6727-OEM 332-6729-OEM 332-6719-OEM 332-6712-OEM 332-6711-OEM 332-6728-OEM
76018-570EA 1005.47 USD
76018-570 75805-440 75805-522 75805-524 75805-442 76018-578 76018-580 76018-730 76018-572 76018-576 75805-520 75805-438 76018-728 76018-574
EZwaste® XL 75 L Closed System for HPLC Solvent Waste, HDPE Reusable Carboy, Foxx Life Sciences
Filters Liquid Process Filters
This 75 L system allows for the safe and effective collection of waste in the laboratory. The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of ext.Ø tubing port configurations
  • Hose Barb option in ¹/₄ or ³/₈"
The EZwaste® XL Solvent Waste System is designed to be an affordable solution for containing liquid chromatography waste.
Each EZwaste® Solvent Waste System comes with two 120 mm VersaCap® (one open top for waste collection and one closed top for safe transport), fittings, plugs for unused ports, and a chemical exhaust filter. Depending on application, carbon exhaust filter life cycle may vary.
Tube fittings secure waste tubes to carboy, providing the closed system necessary to prevent spills. A 90° adapter is included for maximum storage flexibility. A replaceable sulfur cured EPDM gasket is included for high chemical compatibility against aggressive organic solvents.
Each system is manufactured to safely collect waste from up to two HPLC machines. This system features an activated carbon filter, which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers, and provides an easy mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste System also features a closed-loop design which protects against leaks and spills, improving overall lab safety.