EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Supplier: Foxx Life Sciences
330-0427-OEM 330-0411-OEM 330-0413-VWR 330-0429-OEM 330-0418-VWR 330-0426-OEM 330-0420-OEM 330-0425-OEM 330-0428-OEM 330-0431-OEM 330-0430-OEM 330-0421-VWR 330-0412-VWR 330-0419-VWR
76018-506EA 378.89 USD
76018-506 76018-458 76018-508 10827-858 10827-856 10827-862 76018-510 10827-864 76018-464 76018-472 76018-462 76018-470 10827-860 76018-460
EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Closures Bottle Closures
This cap system allows for the safe and effective collection of waste in the laboratory
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb option in 1/4" or 3/8"
The EZwaste® Solvent Waste System is designed to be an affordable solution forcontaining liquid chromatography waste. Each system is manufactured to safely collect waste from one or more HPLC systems while the carbon exhaust filters improve air quality by capturing potentially harmful vapors emitted from solvents. Each cap configuration includes an 83 mm VersaCap®, open top for waste collection, 6 ports with a variety of O.D. configurations for waste tubes, one port for a carbon exhaust filter, one carbon exhaust filter, and plugs for unused ports. Tube fittings secure waste tubes to the carboy, providing the closed system necessary to prevent spills (plugs included for unused ports). A 90° adapter is included for maximum storage flexibility.
The safety and ease of use for laboratory technicians is a high priority. The closed-loop design prevents inhalation of volatile organic componds.