VWR® Volumetric Flask, Amber, Narrow Neck, Class A, Unserialized
Supplier: VWR, part of Avantor
VWR®
134.234.05A 134.234.00A 134.232.00BA 134.232.06A 134.232.02A 134.232.03A 134.478.00A 134.232.04A 134.478.02A 134.478.06A 134.232.00A 134.234.00BA 134.234.03A 134.232.01A 134.234.04A 134.234.06A 134.234.02A 134.232.08A 134.478.04A 134.478.01A 134.234.07A 134.232.05A 134.478.00BA 134.234.01A 134.478.05A 134.478.07A 134.478.08A 134.234.08A 134.232.07A
76003-606CS 155.03 USD
76003-606 76003-824 76003-604 76003-768 76003-608 76003-954 76003-656 76003-756 76003-612 76003-610 76003-754 76003-614 76003-758 76003-658 76003-818 76003-816 76017-938 76003-784 76003-762 76003-662 76003-760 76003-782 76003-822 76003-602 76003-766 76003-820 76003-764 76003-600 76003-660
VWR® Volumetric Flask, Amber, Narrow Neck, Class A, Unserialized
Flasks Volumetric Flasks
VWR® flasks are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Manufactured per ASTM E 288 Class specification
  • White diamond enamel graduation lines
  • Calibrated 'to Contain'
VWR® Volumetric Flasks are used for precise dilutions and preparations of standard solutions. These flat bottom flasks have a pear-shaped lower area with a long neck. Calibration and tolerances are in compliance to standards: ASTM E288, ASTM E542, and ASTM E792.
Certificaciones: Flasks are supplied with Batch certificates.