VWR® Round Bottom Flask, 3 Necks, Angled
Supplier: VWR, part of Avantor
VWR®
060.402.28 060.402.27 060.402.36 060.402.25 060.402.37A 060.402.30A 060.402.29 060.402.21 060.402.30 060.402.17 060.402.20 060.402.24A 060.402.23 060.402.32 060.402.26 060.402.24
76012-820CS 136.27 USD
76012-820 76008-630 76008-640 76012-812 76008-632 76008-642 76012-814 76008-634 76012-816 76008-644 76008-636 76008-646 76012-818 76008-638 76008-648 76008-628
VWR® Round Bottom Flask, 3 Necks, Angled
Flasks Round Bottom Flasks
Durable round bottom flasks with 3 necks at an angle.
  • Good temperature resistance
  • Chemically resistant
  • Uniform wall thickness
Manufactured from ASTM E438 Type 1 Class A borosilicate 33 expansion glass. The flasks feature uniform wall thickness which makes them ideal for heating.