VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Serialized
Supplier: VWR, part of Avantor
VWR®
139.454.07A 139.454.05A 139.454.04A 139.454.03A 139.454.02A 139.454.08A 139.454.06A 139.454.01A
76004-044CS 403.3 USD
76004-044 76004-034 76004-036 76004-038 76004-040 76004-030 76004-042 76004-032
VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Serialized
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Class A serialized measuring cylinders with a hexagonal base
  • Calibrated 'to Contain'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certificaciones: Cylinders are supplied with individual work certificates.